Skip to Content
Merck
All Photos(1)

Key Documents

215228

Sigma-Aldrich

Bis(2-nitrophenyl) disulfide

99%

Synonym(s):

2-Nitrophenyl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2NC6H4)2S2
CAS Number:
Molecular Weight:
308.33
Beilstein:
1016770
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

194-197 °C (lit.)

functional group

disulfide
nitro

SMILES string

[O-][N+](=O)c1ccccc1SSc2ccccc2[N+]([O-])=O

InChI

1S/C12H8N2O4S2/c15-13(16)9-5-1-3-7-11(9)19-20-12-8-4-2-6-10(12)14(17)18/h1-8H

InChI key

NXCKJENHTITELM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

PVC membrane electrode based on bis(2-nitrophenyl)disulfide carrier acts as sensor for zinc ions. Bis(2-nitrophenyl) disulphide undergoes mechanochemical grinding with bis(4-chlorophenyl)disulfide in the presence of 1,8-diazabicyclo[5.4.0]undec-7-ene catalyst via metathesis reaction to yield non-symmetrical heterodimer 4-chlorophenyl-2′-nitrophenyl disulfide.

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M B Gholivand et al.
Talanta, 59(2), 399-407 (2008-10-31)
A PVC membrane electrode based on bis(2-nitrophenyl)disulfide carrier exhibits a very good response for Zn(2+) in a wide concentration range (from 2.9x10(-7) to 3.2x10(-2) mol l(-1)) with a slope of 29.9+/-0.4 mV per decade of Zn(2+) concentration. The response time
Szymon Sobczak et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 24(35), 8769-8773 (2018-04-21)
This work describes, for the first time, the application of combined pressure and temperature stimuli in disulfide metathesis reactions. In the system studied, above a pressure of 0.2 GPa, equimolar amounts of symmetric disulfides bis 4-chlorophenyl disulfide [(4-ClPhS)2 ] and bis
Tomislav Friščić
Chemical Society reviews, 41(9), 3493-3510 (2012-03-01)
Mechanochemical reactions effected by milling or grinding are an attractive means to conduct chemical reactions dependent on molecular recognition and to systematically explore different modes of molecular self-assembly. The natural relationship between milling mechanochemistry and supramolecular chemistry arises primarily from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service