Skip to Content
Merck
All Photos(2)

Documents

191779

Sigma-Aldrich

5-Methyl-2-nitrobenzyl chloride

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)CH2Cl
CAS Number:
Molecular Weight:
185.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

41-43 °C (lit.)

SMILES string

Cc1ccc(c(CCl)c1)[N+]([O-])=O

InChI

1S/C8H8ClNO2/c1-6-2-3-8(10(11)12)7(4-6)5-9/h2-4H,5H2,1H3

InChI key

BWPZDLVOJNBEKI-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Luo et al.
Pharmaceutical research, 10(6), 918-923 (1993-06-01)
A number of isoindolo[1,2-b]quinazolines and some benzo[4,5]isoquinolino[1,2-b]quinazolines as structural modification analogues of the antitumor compound batracylin were synthesized and evaluated against HL-60 cell growth and in topoisomerase II-mediated DNA cleavage assays. Of the compounds studied, 10,12-dihydro-7,8-methylenedioxyisoindolo[1,2-b]quinazolin-1 2(10H)-one (1d), 2-amino-10,12-dihydroisoindolo[1,2-b]quinazolin-12(10H)-one (1p)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service