Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

190713

Decanenitrile

Synonym(s):

Caprinitrile

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)8CN
CAS Number:
Molecular Weight:
153.26
Beilstein:
1750910
EC Number:
MDL number:
PubChem Substance ID:

SMILES string

CCCCCCCCCC#N

InChI

1S/C10H19N/c1-2-3-4-5-6-7-8-9-10-11/h2-9H2,1H3

InChI key

HBZDPWBWBJMYRY-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

206.6 °F - Pensky-Martens closed cup

Flash Point(C)

97 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service