Skip to Content
Merck
All Photos(1)

Key Documents

16445

Sigma-Aldrich

Tris(3,3,4,4,5,5,6,6,6-nonafluorohexyl)tin bromide

~90%

Synonym(s):

Bromotris(3,3,4,4,5,5,6,6,6-nonafluorohexyl)stannane, Tris(1H,1H,2H,2H-perfluorohexyl)tin bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12BrF27Sn
CAS Number:
Molecular Weight:
939.86
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

~90%

form

liquid

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)CC[Sn](Br)(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/3C6H4F9.BrH.Sn/c3*1-2-3(7,8)4(9,10)5(11,12)6(13,14)15;;/h3*1-2H2;1H;/q;;;;+1/p-1

InChI key

GHTDTEBNYTUTDL-UHFFFAOYSA-M

Legal Information

Product of Fluorous Technologies, Inc. Protected by U.S. patents 6,156,896; 5,859,247; and 5,777,123

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service