Skip to Content
Merck
All Photos(1)

Key Documents

118311

Sigma-Aldrich

2-Decanol

98%

Synonym(s):

Methyl octyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH(OH)CH3
CAS Number:
Molecular Weight:
158.28
Beilstein:
1719664
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39020226
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.434 (lit.)

bp

211 °C (lit.)

mp

−6-−4 °C (lit.)

density

0.827 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCC(C)O

InChI

1S/C10H22O/c1-3-4-5-6-7-8-9-10(2)11/h10-11H,3-9H2,1-2H3

InChI key

ACUZDYFTRHEKOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Decanol undergoes Ru(OCOCF3)2(CO)(PPh3)2-catalyzed hydrogen elimination reaction.

Application

2-Decanol was used to develop a miniature catalytic reactor for the oxidation of alcohols with O2 in supercritical CO2. It was used to study the substrate spectrum of phytanoyl-CoA hydroxylase with regard to the length of both the acyl chain and the branch at position.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F

Flash Point(C)

85 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salwa Tsouli Sarhir et al.
Journal of food science and technology, 56(8), 3836-3845 (2019-08-16)
Lben is one of the main dairy products in Morocco, which broadly plays a significant role in food industry due to its nutritional, taste, aroma and health features. Aroma is a main quality factor for this kind of fermented dairy
Rongfeng Hu et al.
Cerebral cortex (New York, N.Y. : 1991), 27(6), 3110-3124 (2016-06-03)
Local inhibition by γ-amino butyric acid (GABA)-containing neurons is of vital importance for the operation of sensory cortices. However, the physiological response patterns of cortical GABAergic neurons are poorly understood, especially in the awake condition. Here, we utilized the recently
Seraj A Ansari et al.
Dalton transactions (Cambridge, England : 2003), 45(43), 17216-17222 (2016-11-02)
Isothermal titration calorimetry was employed for the direct measurement of the enthalpy of extraction (ΔH
Highly sustainable catalytic dehydrogenation of alcohols with evolution of hydrogen gas.
Ligthart GBWL, et al.
Tetrahedron Letters, 44(7), 1507-1509 (2003)
Continuous heterogeneous catalytic oxidation of primary and secondary alcohols in scCO2.
Chapman AO, et al.
Green Chemistry, 12(2), 310-315 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service