Skip to Content
Merck
All Photos(4)

Key Documents

101397

Sigma-Aldrich

2,4-Dinitrotoluene

97%

Synonym(s):

2,4-DNT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)2
CAS Number:
Molecular Weight:
182.13
Beilstein:
1912834
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 102.7 °C)

Assay

97%

form

solid

mp

67-70 °C (lit.)

SMILES string

CC1=C([N+]([O-])=O)C=C([N+]([O-])=O)C=C1

InChI

1S/C7H6N2O4/c1-5-2-3-6(8(10)11)4-7(5)9(12)13/h2-4H,1H3

InChI key

RMBFBMJGBANMMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

311.0 °F - closed cup

Flash Point(C)

155.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tania K Naqvi et al.
Journal of hazardous materials, 407, 124353-124353 (2020-11-05)
We demonstrate the detection of dipicolinic acid, (DPA), a biomarker of bacterial spores for Bacillus anthracis, 2,4-Dinitrotoluene (DNT) and picric acid (PA) nitroaromatic hazardous chemicals on ultra-sensitive, reusable femtosecond laser textured Au nanostructures decorated with hierarchical AuNPs as a SERS
Amay J Bandodkar et al.
The Analyst, 138(18), 5288-5295 (2013-07-19)
Increasing security needs require field-deployable, on-the-spot detection tools for the rapid and reliable identification of gunshot residue (GSR) and nitroaromatic explosive compounds. This manuscript presents a simple, all-solid-state, wearable fingertip sensor for the rapid on-site voltammetric screening of GSR and
Tereza Hudcova et al.
Journal of hazardous materials, 192(2), 605-613 (2011-06-15)
The degradation efficiencies of isomeric mononitrotoluenes (2- and 4-NTs) and dinitrotoluenes (2,4-DNT and 2,6-DNT) by either individual bacterial strains (Bacillus cereus NDT4, Pseudomonas putida NDT1, Pseudomonas fluorescens NDT2, and Achromobacter sp. NDT3) or their mixture were compared in submerged batch
Eric J Olson et al.
Journal of the American Chemical Society, 133(32), 12858-12865 (2011-07-09)
Polynitroaromatics are well-known to form anionic σ-complexes (Meisenheimer complexes). The formation of such complexes was assumed in the past to explain the blue color of solutions of 2,4-dinitrotoluene (DNT) and amines. However, this work shows that caution is warranted to
Jingbo Xu et al.
Journal of hazardous materials, 203-204, 299-307 (2012-01-03)
In this study relative condition factor (RCF) and hepatosomatic index (HSI) as well as the available energy reserves of common carp (Cyprinus carpio) by 2,4-DNT semi-static bioassay were determined and linked to effects of enzymes in liver tissues. Fish were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service