Skip to Content
Merck
All Photos(2)

Key Documents

572365

Sigma-Aldrich

Cerium(IV) oxide-samarium doped

nanopowder, contains 15 mol % samarium as dopant

Synonym(s):

SDC, Samarium-doped ceria

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CeO2
Molecular Weight:
172.11
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

form

nanopowder

contains

15 mol % samarium as dopant

composition

Ce0.85Sm0.15O2

reaction suitability

reagent type: catalyst
core: cerium

particle size

<100 nm

application(s)

battery manufacturing

SMILES string

[Sm].O=[Ce]=O

InChI

1S/Ce.2O.Sm

InChI key

AQGZISMLQKBRRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cerium(IV) oxide-samarium doped nanopowder (15% mol samarium doped) (SDC) is an electrolytic material with a high oxygen ion conductivity. Samarium ion acts as an aliovalent cation that can be incorporated into the host lattice to increase the oxygen vacancy concentration and improve the oxide ion conduction in cerium oxide. It can be synthesized by reverse strike co-precipitation technique.
Samarium doped ceria may be used as an electrolyte in solid oxide fuel cells (SOFCs). Electrical conductivity as a function of temperature of doped ceria was investigated in a study. Relation of dopant type, oxygen vacancy ordering and ionic conductivity was investigated by transmission electron microscopy and electron energy loss spectroscopy.

Application

Cerium(IV) oxide-samarium doped can be used in the fabrication of solid oxide fuel cells (SOFCs), which can further be used as environmentally-friendly sources of alternative energy.

Packaging

Packaged in glass bottles

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Intermediate-temperature electrochemical performance of a polycrystalline PrBaCo2O5+ δ cathode on samarium-doped ceria electrolyte.
Chen D, et al.
Journal of Power Sources, 188(1), 96-105 (2009)
Stephan Immenschuh et al.
Transplantation, 99(1), 56-62 (2014-08-15)
Treatment of patients with antibody-mediated rejection (AMR) after kidney transplantation by rituximab and plasmapheresis is ambiguous. Because of its unknown efficiency and serious side effects, biomarkers, which are predictive for responsiveness to this treatment in AMR patients, are required. Twenty
Fabrication and electrochemical properties of epitaxial samarium-doped ceria films on SrTiO3-buffered MgO substrates
Sanna S, et al.
Advances in Functional Materials, 19(11), 1713-1719 (2009)
Jun Zhang et al.
Scientific reports, 4, 7082-7082 (2014-11-19)
In this work, redox-active Mn or Cr is introduced to the B site of redox stable perovskite Sr(0.95)Ti(0.9)Nb(0.1)O3.00 to create oxygen vacancies in situ after reduction for high-temperature CO2 electrolysis. Combined analysis using X-ray diffraction, X-ray photoelectron spectroscopy, transmission electron
Harsharan K Singh et al.
Transplantation, 99(3), 609-615 (2014-08-20)
A qualitative highly predictive urinary test for polyomavirus nephropathy (PVN) is the PV-Haufen test. This article evaluates whether a quantitative PV-Haufen analysis, that is, the number of PV-Haufen shed per milliliter urine, predicts PVN disease grades and the severity of

Articles

As with all types of fuel cells, a Solid Oxide Fuel Cell (SOFC) is capable of efficiently transforming chemical energy into electrical energy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service