Separation Options with Sephacryl®
Five Sephacryl® HR chromatography media are available as prepacked columns and in bulk packs (Table 1).
Product | Fractionation range, Mr (globular proteins) | Fractionation range, Mr (dextrans) | Sample loading capacity* | Maximum pressure drop over the packed bed | Recommended operation flow† |
---|---|---|---|---|---|
HiPrep™ 16/60 Sephacryl® S-100 HR | 1 × 103 to 1 × 105 | ≤ 5 mL | 0.15 MPa, 1.5 bar,21 psi | 0.5 mL/min | |
HiPrep™ 26/60Sephacryl® S-100 HR | 1 × 103 to 1 × 105 | ≤ 13 mL | 0.15 MPa, 1.5 bar,21 psi | 1.3 mL/min | |
Sephacryl® S-100 HR(Bulk medium) | 1 × 103 to 1 × 105 | 0.5% to 4% of total column volume | Column-dependent | 10 to 35 cm/h | |
HiPrep™ 16/60Sephacryl® S-200 HR | 5 × 103 to 2.5 × 105 | 1 × 103 to 8 × 104 | ≤ 5 mL | 0.15 MPa, 1.5 bar,21 psi | 0.5 mL/min |
HiPrep™ 26/60Sephacryl® S-200 HR | 5 × 103 to 2.5 × 105 | 1 × 103 to 8 × 104 | ≤ 13 mL | 0.15 MPa, 1.5 bar,21 psi | 1.3 mL/min |
Sephacryl® S-200 HR (Bulk medium) | 5 × 103 to 2.5 × 105 | 1 × 103 to 8 × 104 | 0.5% to 4% of total column volume | Column-dependent | 10 to 35 cm/h |
HiPrep™ 16/60Sephacryl® S-300 HR | 1 × 104 to 1.5 × 106 | 2 × 103 to 4 × 105 | ≤ 5 mL | 0.15 MPa, 1.5 bar,21 psi | 0.5 mL/min |
HiPrep™ 26/60Sephacryl® S-300 HR | 1 × 104 to 1.5 × 106 | 2 × 103 to 4 × 105 | ≤ 13 mL | 0.15 MPa, 1.5 bar,21 psi | 1.3 mL/min |
Sephacryl® S-300 HR(Bulk medium) | 1 × 104 to 1.5 × 106 | 2 × 103 to 4 × 105 | 0.5% to 4% of total column volume | Column-dependent | 10 to 35 cm/h |
HiPrep™ 16/60Sephacryl® S-400 HR | 2 × 104 to 8 × 106 | 1 × 104 to 2 × 106 | ≤ 5 mL | 0.15 MPa, 1.5 bar,21 psi | 0.5 mL/min |
HiPrep™ 26/60Sephacryl® S-400 HR | 2 × 104 to 8 × 106 | 1 × 104 to 2 × 106 | ≤ 13 mL | 0.15 MPa, 1.5 bar,21 psi | 1.3 mL/min |
Sephacryl® S-400 HR(Bulk medium) | 2 × 104 to 8 × 106 | 1 × 104 to 2 × 106 | 0.5% to 4% of total column volume | Column-dependent | 10 to 35 cm/h |
HiPrep™ 16/60Sephacryl® S-500 HR | 4 × 104 to 2 × 107 | ≤ 5 mL | 0.15 MPa, 1.5 bar,21 psi | 0.5 mL/min | |
HiPrep™ 26/60Sephacryl® S-500 HR | 4 × 104 to 2 × 107 | ≤ 13 mL | 0.15 MPa, 1.5 bar,21 psi | 1.3 mL/min | |
Sephacryl® S-500 HR(Bulk medium) | 4 × 104 to 2 × 107 | 0.5% to 4% of total column volume | Column-dependent | 10 to 35 cm/h | |
Sephacryl® S-1000 SF(Superfine,bulk medium) | 0.5% to 4% of total column volume | not determined | 2 to 30 cm/h |
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch