Merck
HomeProtein ExpressionPhospholipase A2
Phospholipase A2
Phospholipase A2 (PLA2) designates a class of enzymes that hydrolyze the sn-2 ester of glycerophospholipids to produce a fatty acid and a lysophospholipid. It has become clear that some of these enzymes liberate arachidonic acid in mammalian cells for the biosynthesis of eicosanoids, and thus there has been considerable interest in developing PLA2 inhibitors. Based on amino acid sequences, there are now more than 12 distinct groups of mammalian PLA2s, as well as many non-mammalian forms, all of which have been classified into 14 distinct groups with many subgroups.
Since naturally occurring phospholipids have virtually no solubility in water, PLA2 must bind to the lipid-water interface to access its substrate, and it is clear that catalysis occurs at the interface (interfacial catalysis). One has to be careful with the interpretation of kinetic data including inhibition data, and in fact many of the previously reported PLA2 inhibitors work by non-specific mechanisms. One of the problems is that the substrate forms an interface, and the inhibitor can potentially partition into the membrane phase. Thus, inhibition of a PLA2 by micromolar amounts of a compound using micromolar amounts of substrate may occur because a large fraction of the interface could be occupied by inhibitor. This may change the physical nature of the interface, causing the enzyme to desorb from the membrane into the aqueous phase, resulting in non-specific inhibition. Inhibitors that work by this non-specific mechanism are clearly not useful for studying the role of PLA2s in complex cellular processes. Among those inhibitors that bind tightly to the active site of PLA2s, and thus operate by a specific mechanism, the issue of PLA2 group specificity is important.
To date, 10 groups of mammalian secreted PLA2s, known as s-PLA2s, have been identified. Group IB PLA2, also known as pancreatic PLA2, is found not only as part of the digestive fluid where it functions to hydrolyze dietary phospholipids, but also in non-digestive tissues including spleen where it has unknown functions. Group IIA PLA2 was the first non-pancreatic mammalian PLA2 to be identified as a component of synovial fluid and platelets. This enzyme is pro-inflammatory, displays potent bactericial properties and is a target for the development of anti-inflammatory agents including anti-sepsis agents. Groups IIC, IID, IIE, IIF, III, X, XIIA and XIIB secreted PLA2 were discovered by recombinant DNA techniques. The group IIC gene is functional in mice but occurs as a pseudogene in humans. Group XIIB is best designated as a secreted PLA2-like protein since it has a natural mutation of a key catalytic residue that renders this protein devoid of phospholipase activity. The function of these recently discovered secreted PLA2s are unknown. Group V PLA2, also discovered at the DNA level, is an active enzyme secreted from macrophages and probably a variety of other cells. Recent gene disruption studies implicate a role of this enzyme in arachidonic acid release in stimulated macrophages. All of these secreted PLA2s have similar size, three-dimensional structure, and active site residues (except for group XIIB as noted above). They require submillimolar amounts of calcium for catalytic activity.
Mammalian cells also contain two intracellular enzymes that act on long-chain phospholipids. Group IVA PLA2 translocates from the cytosol to internal membranes in response to micromolar calcium, and shows specificity for arachidonyl-containing phospholipids. A wide variety of studies have shown that this enzyme, also known as cPLA2a, releases arachidonic acid from membrane phospholipids for the biosynthesis of eicosanoids. Recent paralogs of cPLA2a have been identified in the genome, but their functions are not known. It has been proposed that the calcium-independent group VI PLA2 may be involved in phospholipid remodeling, insulin secretion from b cells and in stores-operated calcium entry. Groups VII and VIII PLA2s are highly specific for phospholipids with short sn-2 chains and are thought to terminate the action of platelet activation factor by hydrolyzing the sn-2 ester and to act on oxidized phospholipids.
Table 1 and Table 2 contain Accepted modulators and additional information. For a list of additional products, see the "Similar Products" section below.
GroupIB
(P8913P6534)a
IIA, IIC, IID, IIE and IIFIIIIVV
SourcesbMammalsMammals
(IIC gene is functional in mice but a pseudogene in humans)
Mammals
Bee and wasp venoms
MammalsMammals
Secreted or CytosolicSecretedSecretedSecretedCytosolicSecreted
Structural Information133 aa (human)
Multiple disulfides
~130 aa
Multiple disulfides
IIF has a C-terminal extension
Mammalian enzyme has a ~16 kDa group III PLA2 domain and long N- and C-terminal extensions749 aa (human) α-paralog
Several new paralogs have been recently identified
118 aa (human)
Multiple disulfides
Molecular Weight (kDa)13 - 1513 - 1755 (mammalian)
18 (venom)
80-85 (α-paralog)14
CofactorµM – mM Ca2+ required for active site substrate binding and catalysiseµM – mM Ca2+ required for active site substrate binding and catalysiseµM – mM Ca2+ required for active site substrate binding and catalysiseSub to low mM Ca2+ required for membrane binding
γ-paralog does not require Ca2+
µM – mM Ca2+ required for active site substrate binding and catalysise
Tight Binding InhibitorscHK series sn-2-amide phospholipids
Indole analogsf
HK series sn-2-amide phospholipids
Indole analogse
HK series sn-2-amide phospholipids
Indole analogse
HK series sn-2-amide phospholipids
Indole analogsf
HK series sn-2-amide phospholipids
Indole analogse
Control CompoundsEnantiomer of HK series
Some Indole analogs
Enantiomer of HK series
Some Indole analogs
Enantiomer of HK series
some Indole analogs
AACOCH3Enantiomer of HK series
Some indole analogs
Inhibitor SpecificityMany group IB inhibitors inhibit other secreted PLA2seMany group II inhibitors inhibit other secreted PLA2seMany group III inhibitors inhibit other secreted PLA2seAACOCF3 (A231) and MAFP also inhibit group VI PLA2SMany group V inhibitors inhibit other secreted PLA2se
Tissue ExpressionbPancreatic juice
Spleen
Lung
Several mammalian tissues
Tissue expression is partially overlapping
Kidney
Heart
Liver
Skeletal muscle
Most mammalian tissuesMacrophages
Lung
Physiological FunctiondDigestion of dietary phospholipidsBactericidal activity (IIA)
Functions of IIC, IID, IIE and IIF are unknown
Not KnownAgonist-triggered arachidonic acid release for eicosanoid and platelet activator factor production (α-isoform)
Functions of other isoforms are unknown
Contributes to arachidonic acid release for eicosanoid production
Augmentation of cPLA2α function
Disease RelevancedMouse IB deficient mice show resistance to obesitySepsis and inflammation, possibly arthritis (IIA)Not KnownInflammation and allergy related to lipid mediator productionNot Known
Table 1.Accepted modulators and additional information
GroupVI
VIIVIIIXXIIA
SourcesbMammalsHuman plasmaBovine brainMammalsMammals
Secreted or CytosolicCytosolicSecretedCytosolicSecretedSecreted
Structural InformationiPLA2a isoform is 752 aa (human)
Several isoforms and splice forms are known
420 aa (human)230 aa (bovine γ-subunit)123 aa (human)167 aa (human)
Multiple disulfides
Molecular Weight (kDa)iPLA2a isoform 80 - 8545291419
CofactorNone
May be augmented by ATP
Not Known
Not Known
µM – mM Ca2+ required for active site substrate binding and catalysiseµM – mM Ca2+ required for active site substrate binding and catalysise
Tight Binding InhibitorscBEL (B1552)
AACOCF3 (A231)
MAFP
Patented inhibitors from GSKNot KnownAACOCF3 (A231)
MAFP
Pyrrophenone and related compounds
Not Known
Control CompoundsEnantiomers BEL (B1552) have distinct specificity for the various iPLA2 isoformsNot Known
Not KnownEnantiomer of HK series
Some indole analogs
Not Known
Inhibitor SpecificityAACOCF3 (A231) and MAFP also inhibit cPLANot Known
Not KnownMany group IB inhibitors inhibit other secreted PLA2seNot Known
Tissue ExpressionbExpressed in many mammalian tissuesHuman plasma
An isoform is expressed in mammalian cytosol
BrainSeveral mammalian tissuesSeveral mammalian tissues
Physiological FunctiondStores operated calcium entry
Insulin secretion
Phospholipid acyl chain remodeling
Serum form degrades platelet activating factor and phospholipids with oxidized fatty acyl chainsNot KnownNot KnownNot Known
Disease RelevancedNot Known
Not Known
Mutations cause Miller-Dieker lissencephalyNot KnownNot Known
Table 2.Accepted modulators and additional information
Footnotes
a) Not included in the table are a variety of other PLA2s from non-mammalian sources including PLA2s in snake and insect venoms, for example the group IA enzymes in cobra and krait venoms (P7778), rattlesnakes and bee venom (P9279).
b) The species and tissue distribution is only a partial listing.
c) Only those inhibitors that have been shown to bind specifically to the active site of the PLA2s have been listed (see text for more discussion); and the list is not necessarily comprehensive.
d) The list of physiological functions and disease relevance is only partial as the functions of PLA2s is under active investigation and is still unresolved in many cases.
e) See, Singer et al., J Biol. Chem., 277, 48535-49 (2002).
f) See, Smart, B.P., et al., Bioorg. Med. Chem., 12, 1737-1749 (2004).
Abbreviations
AACH(OH)CF3: 2-Hydroxy-1,1,1,-trifluoro-6,9,12,15-heneicosatetraene
AACOCF3: 2-oxo-1,1,1-Trifluoro-6,9-12,15-heneicosatetraene
AACOCH3: 2-oxo-6,9,12,15-Heneicosatetetraene
BEL: (E)-6-(Bromomethylene)tetrahydro-3-(1-naphthalenyl)-2H-pyran-2-one
MAFP: 4,7,10,13-Nonadecatetraenyl fluorophosphonic acid methyl ester
SB-222657: N-[6-(4-Chlorophenyl)hexyl]-2-oxo-4-[(S)-(phenylmethyl)sulfinyl]-1-azetidineacetamide
SB-223777: N-[6-(4-Chlorophenyl)hexyl]-2-oxo-4-[(R)-(phenylmethyl)sulfinyl]-1-azetidineacetamide
Similar Products
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
Balsinde J, Winstead MV, Dennis EA. 2002. Phospholipase A2 regulation of arachidonic acid mobilization. 531(1):2-6. https://doi.org/10.1016/s0014-5793(02)03413-0
2.
Bonventre JV, Sapirstein A. 2002. Group IV Cytosolic Phospholipase A2(Pla2) Function:Insights from the Knockout Mouse.25-31. https://doi.org/10.1007/978-1-4615-0193-0_5
3.
Cao J, Burke JE, Dennis EA. 2013. Using Hydrogen/Deuterium Exchange Mass Spectrometry to Define the Specific Interactions of the Phospholipase A2Superfamily with Lipid Substrates, Inhibitors, and Membranes. J. Biol. Chem.. 288(3):1806-1813. https://doi.org/10.1074/jbc.r112.421909
4.
Dan P, Rosenblat G, Yedgar S. 2012. Phospholipase A2 activities in skin physiology and pathology. European Journal of Pharmacology. 691(1-3):1-8. https://doi.org/10.1016/j.ejphar.2012.07.023
5.
Dennis EA, Cao J, Hsu Y, Magrioti V, Kokotos G. 2011. Phospholipase A2Enzymes: Physical Structure, Biological Function, Disease Implication, Chemical Inhibition, and Therapeutic Intervention. Chem. Rev.. 111(10):6130-6185. https://doi.org/10.1021/cr200085w
6.
Gelb MH, Jain MK, Berg OG. 1994. Inhibition of phospholipase A 2. FASEB j.. 8(12):916-924. https://doi.org/10.1096/fasebj.8.12.8088457
7.
Gentile MT, Reccia MG, Sorrentino PP, Vitale E, Sorrentino G, Puca AA, Colucci-D?Amato L. 2012. Role of Cytosolic Calcium-Dependent Phospholipase A2 in Alzheimer's Disease Pathogenesis. Mol Neurobiol. 45(3):596-604. https://doi.org/10.1007/s12035-012-8279-4
8.
Leslie CC. 1997. Properties and Regulation of Cytosolic Phospholipase A2. J. Biol. Chem.. 272(27):16709-16712. https://doi.org/10.1074/jbc.272.27.16709
9.
Ma Z, Turk J. 2001. The molecular biology of the group VIA Ca2+-independent phospholipase A2.1-33. https://doi.org/10.1016/s0079-6603(01)67023-5
10.
Murakami M, Kudo I. 2001. Diversity and regulatory functions of mammalian secretory phospholipase A2s.163-194. https://doi.org/10.1016/s0065-2776(01)77017-4
11.
P. Samy R, Gopalakrishnakone P, G. Stiles B, S. Girish K, N. Swamy S, Hemshekhar M, S. Tan K, G. Rowan E, Sethi G, T.K. Chow V. 2012. Snake Venom Phospholipases A2: A Novel Tool Against Bacterial Diseases. Current Medicinal Chemistry. 19(36):6150-6162. https://doi.org/10.2174/092986712804485791
12.
Satake Y, Diaz BL, Balestrieri B, Lam BK, Kanaoka Y, Grusby MJ, Arm JP. 2004. Role of Group V Phospholipase A2in Zymosan-induced Eicosanoid Generation and Vascular Permeability Revealed by Targeted Gene Disruption. J. Biol. Chem.. 279(16):16488-16494. https://doi.org/10.1074/jbc.m313748200
13.
Singer AG, Ghomashchi F, Le Calvez C, Bollinger J, Bezzine S, Rouault M, Sadilek M, Nguyen E, Lazdunski M, Lambeau G, et al. 2002. Interfacial Kinetic and Binding Properties of the Complete Set of Human and Mouse Groups I, II, V, X, and XII Secreted Phospholipases A2. J. Biol. Chem.. 277(50):48535-48549. https://doi.org/10.1074/jbc.m205855200
14.
Six DA, Dennis EA. 2000. The expanding superfamily of phospholipase A2 enzymes: classification and characterization. Biochimica et Biophysica Acta (BBA) - Molecular and Cell Biology of Lipids. 1488(1-2):1-19. https://doi.org/10.1016/s1388-1981(00)00105-0
15.
Smani T, Zakharov SI, Csutora P, Leno E, Trepakova ES, Bolotina VM. 2004. A novel mechanism for the store-operated calcium influx pathway. Nat Cell Biol. 6(2):113-120. https://doi.org/10.1038/ncb1089
16.
Uozumi N, Kume K, Nagase T, Nakatani N, Ishii S, Tashiro F, Komagata Y, Maki K, Ikuta K, Ouchi Y, et al. 1997. Role of cytosolic phospholipase A2 in allergic response and parturition. Nature. 390(6660):618-622. https://doi.org/10.1038/37622
17.
Valentin E, Lambeau G. 2000. Increasing molecular diversity of secreted phospholipases A2 and their receptors and binding proteins. Biochimica et Biophysica Acta (BBA) - Molecular and Cell Biology of Lipids. 1488(1-2):59-70. https://doi.org/10.1016/s1388-1981(00)00110-4