Merck
HomeProtein ExpressionGlutamate Receptors (Ion Channel Family)
Glutamate Receptors (Ion Channel Family)
The ion channel family of glutamate receptors ( ionotropic; glutamate receptors or iGluR) comprises three major subtypes based on pharmacology and protein structure. 
The N-methyl-D-aspartate (NMDA) subtype is a hetero-oligomer consisting of an NR1 subunit combined with one or more NR2 (A-D) subunits and a third subunit, NR3 (A,B). The receptor has two amino acid recognition sites, one for glutamate and one for glycine, both of which must be occupied to promote channel opening. Antagonists have been discovered which selectively compete for either the glutamate or glycine site, and these act as functional receptor antagonists. The channel is permeable to cations, including calcium, and is blocked by magnesium at membrane potentials close to resting, endowing a voltage dependence to this ligand-gated ion channel which is important for its physiological role as a " conditional" receptor. A variety of drugs have been identified which block the channel selectively. Other sites exist on the receptor through which polyamines, zinc, protons and oxidizing/reducing agents influence receptor function. Several classes of compounds have been identified that interact selectively with NMDA receptors containing NR2B, providing "subunit-selective" antagonists. The NMDA receptor is recognized as part of a large complex of cell surface proteins, receptors and intracellular mediators at the post-synaptic density, which interact to regulate excitatory neurotransmission and synaptic plasticity.
The α-amino-3-hydroxy-5-methylisoxazole-4-propionic acid (AMPA) subtype is a hetero-oligomer formed from combinations of iGluR1-4. Selective agonists and competitive antagonists acting at the glutamate recognition site have been useful for defining the physiological and pathophysiological roles played by the receptor. Allosteric sites on the receptor mediate the effects of the AMPAkines that potentiate the response to glutamate while GYKI 52466 and related compounds act as non-competitive inhibitors. The ion channel is cation-permeable, but calcium permeability is regulated by an RNA editing mechanism in the iGluR2 subunit. Joro spider toxin blocks the channel of AMPA receptors that do not contain the GluR2 subunit.
The kainate subtype consists of hetero-oligomers, comprising the iGluR5-7 and KA1 and KA2 subunits. Kainate itself and the AMPA analog ATPA have been used as selective agonists and recently, several competitive antagonists with selectivity for kainate receptors, particularly those containing iGluR5 subunits, have been identified.
The physiological and pathophysiological roles of ionotropic glutamate receptors have been extensively studied with both molecular and pharmacological approaches. NMDA receptors are post-synaptic and play important roles in plasticity in the developing and mature CNS and post-synaptic AMPA receptors mediate chemical transmission at the majority of fast excitatory synapses in the CNS. Both NMDA and AMPA receptors play key roles in certain forms of synaptic plasticity such as long-term potentiation and depression, phenomena that may underlie physiological processes such as learning and memory. Due to their voltage dependence and calcium permeability, NMDA receptors play a key role in the initiation of synaptic plasticity and the expression of these phenomena are due to alterations in the cell surface expression of AMPA receptors. The role of kainate receptors is less clear, but in the hippocampus and spinal cord, there is evidence for a presynaptic location and an influence on transmitter release. The evidence that NMDA receptors are involved in neurodegenerative processes led to numerous unsuccessful clinical trials in stroke and head trauma with drugs that antagonize the receptor through binding to the glutamate, glycine and allosteric sites. AMPA receptor antagonists also have neuroprotective and anticonvulsant properties in animal studies. Memantine, which is a low affinity NMDA receptor channel blocker has shown cognitive improvements in Alzheimer's patients. NMDA receptor antagonists have also shown beneficial effects to alleviate the motor dysfunction in Parkinson' s disease and relieve pain in animal models. The beneficial effects of AMPAkines in animal models of cognitive impairment and psychosis have prompted clinical studies in these areas. Kainate receptor antagonists have shown positive effects in animal and human models of pain and migraine.
The Tables below contains accepted modulators and additional information. For a list of additional products, see the Materials section below.
Currently Accepted NameGlutamate siteNMDA Glycine siteOther
Alternate Name  Quisqualate
Structural InformationNR1 (920 aa human)
NR2A (1464 aa human)
NR2B (1484 aa human)
NR2C (1233 aa human)
NR2D (1329 aa rat)
NR3A (1115 aa rat)
NR3B (1003 aa mouse)
Not KnownNot Known
Subtype Selective AgonistsN-Methyl-D-aspartic acid (M3262)
Quinolinic acid (P63204)
Glycine (G7126)
D-Serine (S4250)
R(+)-HA-966 (H130) (partial)
Cyclothiazided (C9847)
LY503430d
Subtype Selective AntagonistsD(–)-AP-5 (A8054)
D(–)-AP-7
CGS19755
CGP37849 (SML0050)
LY382884 (GluR5)
(±)-CPP (C104)
D-CPP
D-CPPene
EAA-090 (SML0206)
7-Chlorokynurenic acid
5,7-Dichlorokynurenic acid (D138)
MNQX
L-689,560
L-701,324 (L0258)
GV 150526
Ro 25-6981 (R7150) (NR2B)
Ro 8-4304 (R8900) (NR2B)
CP 101,606 (SML0053) (NR2B)
Ifenprodil (I2892) (NR2B)
SPD-502
Channel BlockersMK-801 (Dizocilpine) (M107)
Phencyclidine (PCP) (P3029)
CNS-1102 (Cerestat) (C4238)
Ketamine (K2753)
Memantine (M9292)
Not KnownNot Known
Channel PermeabilityIntrinsic ion channel (Na+/K+/Ca2+)Not KnownNot Known
Radioligands of Choice[3H]-CPP
[3H]-L-Glutamate
[3H]-5,7-Dichlorokynurenate
[3H]-L-689,560
[3H]-MK-801 (channel)
[3H]-Ro 25-6981 (NR2B)
Tissue Expression Receptors are widespread in the CNS 
Physiological FunctionEach class of receptors is involved in fast synaptic transmission, and neuronal plasticity in the developing and adult CNS; Kainate receptors have presynaptic role
Disease Relevance Stroke, Alzheimer's disease, Parkinson's
disease, schizophrenia, pain, epilepsy
Table 1.
Currently Accepted NameAMPAKainate
Alternate Name  
Structural InformationiGluR1 (889 aa human)
iGluR2 (883 aa human)
iGluR3 (894 aa human)
iGluR4 (881 aa rat)
iGluR5 (978 aa human)
iGluR6 (877 aa rat)
iGluR7 (919 aa human)
KA1 (956 aa human)
KA2 (962 aa human)
Subtype Selective AgonistsAMPA (A0326)
S(–€“)-5-Fluorowillardiine
CX-614d
Cyclothiazided (C9847)
LY503430d
Kainic acid (K0250)
Domoic acid (D6152)
4-Methylglutamate (G137)
Subtype Selective AntagonistsNBQX (N183)
GYKI 52466c (G119)
GYKI 53655c (G7798)
CNQX (C239)
DNQX (D0540)
YM90K
LY294486
Ro 48-8587
SPD-502
CNQX (C239)
DNQX (D0540)
NS 102b (N179)
LY293558 (iGluR5)
NS3763 (N1162) (iGluR5)
Channel BlockersJoro Spider Toxin
Not Known
Channel PermeabilityIntrinsic ion channel (Na+/K+/Ca2+)Intrinsic ion channel (Na+/K+/Ca2+)
Radioligands of Choice[3H]-AMPA
[3H]-Ro 48-8587
[3H]-Kainic acid
[3H]-NBQX
Tissue ExpressionReceptors are widespread in the CNSReceptors are more discretely localized in areas of
cortex, hippocampus, thalamus and spinal cord
Physiological FunctionEach class of receptors is involved in fast synaptic transmission, and neuronal plasticity in the developing and adult CNS; Kainate receptors have presynaptic role
Disease RelevanceEpilepsy, stroke, Alzheimer'€™s disease, schizophreniaPain, epilepsy
Table 2.
Footnotes
a) Ion channel family is also referred to as ionotropic.
b) Selectively inhibits low affinity [3H]-kainate binding.
c) Non-competitive antagonist.
d) Allosteric potentiator.
Abbreviations
AMPA: α-Amino-3-hydroxy-5-methylisoxazole-4-propionic acid
AP-5: 2-Amino-5-phosphonopentanoic acid
AP-7: 2-Amino-7-phosphonoheptanoic acid
ATPA: (RS)-2-Amino-3-(3-hydroxy-5-tert-butylisoxazol-4-yl)propanoic acid
D-CCPene: D-3-(2-Carboxypiperazin-4-yl)-propyl-1-phosphonene
CGP37849: D,L-(E)-2-Amino-4-methylphosphono-3-pentanoic acid
CGS19755: 4-Phosphonomethyl-2-piperidinecarboxylic acid (Selfotel)
CNQX: 6-Cyano-7-nitroquinoxaline-2,3-dione
CNS 1102: N-(1-Naphthyl)-N'-(3-ethylphenyl)-N'-methyl-guaniaine HCl
CP 101,606: (1S,2S)-1-(4-Hydroxyphenyl)-2-(4-hydroxy-4-phenylpiperidino)-1-propanol
CPP: 3-(2-Carboxypiperazin-4-yl)-propyl-1-phosphonic acid
CX-614: 2H,3H,6aH-Pyrolidino[2",1"-3' ,2' ]1,3-oxazino[6' ,5' ,-5,4]benzo[e]1,4-dioxan-10-one
DNQX: 6,7-Dinitroquinoxaline-2,3-dione
EAA-090: [2-(8,9-Dioxo-2,6-diazabicyclo[5.2.0]non-1(7)-en-2-yl)-ethyl]phosphonic acid
GV 150526: 3-[2-(Phenylamino)carbonyl]ethenyl-4,6-dichloroindole-2-carboxylic acid
GYKI 52466: 1-(4-Aminophenyl)-4-methyl-7,8-methylenedioxy-5H-2,3-benzodiazepine
GYKI 53655: 1-(4-Aminophenyl)-3-methylcarbamyl-4-methyl-7,8-methylenedioxy-3,4-dihydro-5H-2,3-benzodiazepine
HA-966: 1-Hydroxy-3-aminopyrrolid-2-one
L-689,560: (±)-4-(trans)-2-Carboxy-5,7-dichloro-4-phenylaminocarbonylamino-1,2,3,4-tetrahydroquinoline
L-701,324: 7-Chloro-4-hydroxy-3-(3-phenoxy)phenyl-2(H)-quinolinone
LY293558: (3S,-4aR,6R,8aR)-6-[2-([1Htetrazol-5-yl)ethyl]decahydroisoquinoline-3-carboxylic acid
LY382884: 3S,4aR,6S,8aR-6-((4-Carboxyphenyl)methyl-1,2,3,4,4a,5,6,7,8,8a-decahydroisoquinoline-3-carboxylic acid
LY503430: (R)-4' -[1-Fluoro-1-methyl-2-(propane-2-sulfonylamino)-ethyl]-biphenyl-4-carboxylic acid methylamide
MNQX: 5,7-Dinitro-1,4-dihydro-2,3-quinoxalinedione
NBQX: 2,3-Dihydro-6-nitro-7-sulphamoyl-benzo(f)quinoxaline
NMDA: N-Methyl-D-aspartic acid
NS 102: 5-Nitro-6,7,8,9-tetrahydrobenzo[G]indole-2,3-dione-3-oxime
NS3763: 5-Carboxyl-2,4-dibenzamido-benzoic acid
Ro 25-6981: R-(R*,S*)-α-(4-Hydroxyphenyl)-β-methyl-4-(phenylmethyl)-1-piperidine propanol
Ro 8-4304: 4-{3-[4-(4-Fluorophenyl)-3,6-dihydro-2H-pyridin-1-yl]-2-hydroxy-propoxy}-benzamide
Ro 48-8587: 9-(1H-Imidazol-1-yl)-8-nitro-[1,2,4]triazolo[1,5-c]quinazoline-2,5(3H,6H)-dione
SPD-502: 8-Methyl-5(4-(N,N-dimethylsulfamoyl)phenyl)6,7,8,9,-tetrahydro-1H-pyrrolo[3,2-h]-isoquinoline-2,3-dione-3-O-(4-hydroxybutyrate-2-yl)oxime
YM90K: 6-(1H-Imidazol-1-yl)-7-nitro-2,3(1H,4H)-quinoxalinedione
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
Bowie D. 2012. Redefining the classification of AMPA-selective ionotropic glutamate receptors. 590(1):49-61. https://doi.org/10.1113/jphysiol.2011.221689
2.
Bräuner-Osborne H, Egebjerg J, Nielsen, Madsen U, Krogsgaard-Larsen P. 2000. Ligands for Glutamate Receptors:  Design and Therapeutic Prospects. J. Med. Chem.. 43(14):2609-2645. https://doi.org/10.1021/jm000007r
3.
Christensen JK. 2004. A Mosaic of Functional Kainate Receptors in Hippocampal Interneurons. Journal of Neuroscience. 24(41):8986-8993. https://doi.org/10.1523/jneurosci.2156-04.2004
4.
Dingledine R ea. 1999 . The glutamate receptor ion channels Pharmacol Rev . 51 (1 ):7-61.
5.
Huettner JE. 2003. Kainate receptors and synaptic transmission. Progress in Neurobiology. 70(5):387-407. https://doi.org/10.1016/s0301-0082(03)00122-9
6.
Jackson A, Nicoll R. 2011. The Expanding Social Network of Ionotropic Glutamate Receptors: TARPs and Other Transmembrane Auxiliary Subunits. Neuron. 70(2):178-199. https://doi.org/10.1016/j.neuron.2011.04.007
7.
Kemp JA, McKernan RM. 2002. NMDA receptor pathways as drug targets. Nat Neurosci. 5(S11):1039-1042. https://doi.org/10.1038/nn936
8.
Larsson M. 2009. Ionotropic Glutamate Receptors in Spinal Nociceptive Processing. Mol Neurobiol. 40(3):260-288. https://doi.org/10.1007/s12035-009-8086-8
9.
Loftis JM, Janowsky A. 2003. The N-methyl-d-aspartate receptor subunit NR2B: localization, functional properties, regulation, and clinical implications. Pharmacology & Therapeutics. 97(1):55-85. https://doi.org/10.1016/s0163-7258(02)00302-9
10.
Lynch G. 2004. AMPA receptor modulators as cognitive enhancers. Current Opinion in Pharmacology. 4(1):4-11. https://doi.org/10.1016/j.coph.2003.09.009
11.
Malenka RC, Bear MF. 2004. LTP and LTD. Neuron. 44(1):5-21. https://doi.org/10.1016/j.neuron.2004.09.012
12.
Mayer ML, Armstrong N. 2004. Structure and Function of Glutamate Receptor Ion Channels. Annu. Rev. Physiol.. 66(1):161-181. https://doi.org/10.1146/annurev.physiol.66.050802.084104
13.
E. McCullumsmith R, Hammond J, Funk A, H. Meador-Woodruff J. 2012. Recent Advances in Targeting the Ionotropic Glutamate Receptors in Treating Schizophrenia. CPB. 13(8):1535-1542. https://doi.org/10.2174/138920112800784899
14.
Mobius H, Stoffler A, Graham S. 2004. Memantine hydrochloride: Pharmacological and clinical profile. Drugs Today. 40(8):685. https://doi.org/10.1358/dot.2004.40.8.850471
15.
Rojas A, Dingledine R. 2013. Ionotropic Glutamate Receptors: Regulation by G-Protein-Coupled Receptors. Mol Pharmacol. 83(4):746-752. https://doi.org/10.1124/mol.112.083352
16.
Smolders I, Bortolotto ZA, Clarke VRJ, Warre R, Khan GM, O'Neill MJ, Ornstein PL, Bleakman D, Ogden A, Weiss B, et al. 2002. Antagonists of GLUK5-containing kainate receptors prevent pilocarpine-induced limbic seizures. Nat Neurosci. 5(8):796-804. https://doi.org/10.1038/nn880
17.
Stawski P, Janovjak H, Trauner D. 2010. Pharmacology of ionotropic glutamate receptors: A structural perspective. Bioorganic & Medicinal Chemistry. 18(22):7759-7772. https://doi.org/10.1016/j.bmc.2010.09.012
18.
Tovar KR, Westbrook GL. 2002. Mobile NMDA Receptors at Hippocampal Synapses. Neuron. 34(2):255-264. https://doi.org/10.1016/s0896-6273(02)00658-x