Merck
HomeBiopharmaceutical CharacterizationQuality Attribute Determination of Monoclonal Antibodies
Quality Attribute Determination of Monoclonal Antibodies
Geoffrey Rule, Principal Scientist, Romana Rigger, Senior Scientist, Pegah Jalili, Principal Scientist, Uma Sreenivasan, Director, Kevin Ray, Senior Manager, Stephan Altmaier, Senior Manager
Merck
Article from Analytix Reporter - Issue 12
Abstract
Three step-by-step protocols for the monitoring of critical quality attributes (CQAs) of monoclonal antibodies were developed using adalimumab as an example. Monoclonal antibodies are glycoproteins used as drugs in the treatment of a variety of cancers and immune system disorders. These large biomolecules have complex structures making thorough characterization essential to ensure the safety and efficacy of drugs. Here, we provide an overview of the different workflows that can be used in these molcules intact and reduced mass analysis, primary structure determination (i.e. peptide map), and N-linked glycan analysis to enable an accurate characterization of their CQAs.
Introduction
Increasing importance of monoclonal antibody (mAb) therapeutics has fundamentally changed the pharmaceutical market in recent years. These macromolecules are used in cancer therapies and in combating autoimmune and other diseases. Biomolecules are produced using recombinant techniques in mammalian cell lines, expressing the mAbs in large bioreactors. mAbs are glycoproteins with a molecular weight of approximately 150 kDa. Antibodies are composed of two light chains and two heavy chains (LC and HC, respectively) linked to one another through covalent inter- and intra-chain disulfide bonds. The individual chains are composed of amino acids and can be post-translationally modified (PTM). The different modifications include glycosylation, phosphorylation, methylation, oxidation, and nitrosylation.
A reliable characterization of mAbs is essential to ensure the safety and efficacy of both the innovator and biosimilar drugs.1 Therefore, the critical quality attributes (CQAs) are defined for each protein along with their acceptable ranges. For example, the extent of deamidation, pyroglutamination, oxidation, or formation of lysine variants in mAbs is quantified in relation to their respective native forms.2 Common analytical techniques employed to verify CQAs being within the set limits include capillary zone electrophoresis, isoelectric focusing, ion exchange, size exclusion (SEC), reversed phase (RP) or hydrophilic interaction liquid chromatography (HILIC), coupled with ultraviolet (UV), fluorescence, or mass spectrometric (MS) detection.3,4 Strategies used to aid the analysis include chemical or enzymatic fragmentation of the protein into subunits,5 removal and analysis of glycans, and proteolytic digestion of the protein into smaller peptides followed by their sequencing. A combination of these techniques can also be applied. For the analysis of mAbs from cell culture supernatants, an affinity purification step is often used to prepare samples prior to their analysis.6
Recently, we prepared step-by-step workflows for three approaches commonly used in mAb characterization.7-9 These approaches determine:
  1. the mass of the intact molecule and of the heavy and light chains (intact and middle-up mass analysis),
  2. the amino acid sequence (peptide mapping), and
  3. the glycosylation state (glycan analysis).
Intact mass analysis relates to the measurement of the mass of an intact mAb without its dissociation into subunits. Middle-up experiments are performed after cleaving the mAbs into several large fragments, or subunits, via chemical reduction or proteolytic digestion. This can provide information on the sites of modification.
In peptide mapping, the protein is cleaved into fragments with a protease enzyme, most commonly trypsin. Prior to the enzymatic digestion, it is common to denature the protein to expose internal amino acids. As in middle-up analysis, the protein may be treated chemically to break disulfide bonds linking different portions of the protein. This reduction step is typically followed by alkylation of the exposed free thiols to prevent reformation of disulfide bonds. Minimizing the introduction of modifications to individual amino acids during the analytical stages of peptide mapping is also important. For example, under appropriate conditions, asparagine can be deamidated and methionine can be oxidized, both of which alter the mass of the peptide.
Glycan analysis is a third means of characterizing CQAs of mAb therapeutics. Glycans are polysaccharide chains that are attached to proteins (glycoprotein) or lipids (glycolipid). They exist as an array of different sugar units attached through different linkages and sometimes existing in complex di-, tri-, and tetra-antennary structures. Glycans are typically described as “N-linked,” when attached at the side chain nitrogen of asparagine, or “O-linked”, when linked to the side chain oxygen of serine or threonine. Glycan profiles can affect the stability and bioavailability of mAb therapeutics and can vary with manufacturing conditions. Characterization of glycans is essential to ensure the equivalence of newly released mAb lots to the approved drug.
Glycan analysis is achieved by releasing glycans from the protein by using chemical or enzymatic methods followed by derivatization and fluorescence or mass spectral analysis. Common derivatization agents include 2-aminobenzamide (2-AB), 2-aminobenzoic acid (2-AA), and procainamide. The latter has properties suited to both fluorescence and MS detection. Fluorescent derivatization allows relative quantification of glycan species while mass spectral analysis provides structural confirmation.
Figure 1 illustrates how the three approaches fit into the overall characterization of a mAb, along with a technique for purification of mAbs when necessary. Next, we look at each of the three workflows in a bit more detail.
Overview of analytical techniques for antibody sample preparation and analysis showing intact mass analysis, peptide mapping, and glycan analysis
Figure 1.Overview of analytical techniques for antibody sample preparation and analysis showcased in this work. A: Intact mass analysis. B: Middle-up analysis of mAb after reduction. C: Peptide mapping after denaturation, reduction, alkylation, and tryptic digestion. D: Glycan analysis after denaturation, glycan release and labelling with procainamide.
Intact mass analysis
Non-reducing (intact) and reducing (middle-up) SEC-MS workflows for mAb analysis provide a high- level characterization of the protein structure and subunits.10 Intact analysis yields mass measurement of the entire mAb, without dissociation of the subunits. Use of high resolution, high mass accuracy mass spectrometers to verify molecular weight can be utilized to reveal information about stoichiometry and proteoforms that may exist. Multiple charge state spectra are deconvoluted to allow correlation of observed masses with expected or theoretical masses.
In SEC, analytes enter, or are excluded from, the pores of the chromatographic support based on size. In a mixture of mAb, mAb aggregates, and antibody fragments, large aggregates elute first, followed by the antibody and then the fragments. In a middle-up analysis, HCs can be separated from LCs in this same fashion to identify the glycosylation sites on one or the other chains.
A photodiode array (280 nm) trace, total ion chromatogram, MS spectrum, and deconvoluted MS spectrum of adalimumab are shown in Figures 2 and 3 (see conditions in Table 1). Observed masses of the non-reduced mAb are found to correlate well with the theoretical masses and observed mass error was found to be 0.010% or less. Deconvolution of the spectrum revealed several different glycoforms.
UV photodiode array trace obtained for intact adalimumab during intact mass analysis
Figure 2.UV photodiode array (280 nm, left) and TIC traces (right) of intact adalimumab.
MS spectrum of intact adalimumab obtained during intact mass analysis
Figure 3.MS spectra for intact adalimumab. Left: summed spectrum; Right: deconvoluted spectrum showing several different glycoforms. (MS spectra generated on a QTOF instrument).
SEC Method Conditions
Column:Tosoh TSK Gel SW3000, 30 cm x 2.0 mm, 4 μm particles (821485)
Mobile phase:Acetonitrile/water 30/70 (v/v) containing 0.1% trifluoroacetic acid (TFA)
Isocratic elution:0.1 mL/min
Column temp.:Ambient
Detection:UV 280 nm and MS
Injection:20 µL
Table 1.SEC Method Conditions for intact mass analysis
Peptide Mapping
Peptide mapping attempts to characterize the exact sequence of amino acids in a protein along with any modifications to those amino acids. In this work, sample proteins were digested with trypsin in a low-artifact digestion buffer11 and utilizing filter-assisted sample preparation (FASP).12 The latter makes use of a molecular weight cut-off filter, in microcentrifuge tube format, to separate peptide fragments from whole protein. The low-artifact digestion buffer minimizes amino acid modification through the digestion process to provide a more accurate depiction of the protein being produced.
The digest is injected onto an HPLC column capable of resolving as many of the peptide fragments as possible, including those bearing glycans, prior to MS detection. HPLC columns based on superficially porous particles (SPP) like Ascentis® Express and BIOshell™ columns are suitable to obtain high efficiencies and resolution. Figure 4 shows the chromatographic profile obtained for the model mAb, adalimumab while Table 2 displays the method conditions. Coupling two 15 cm columns provided greater separation of peptides across the run as well as slightly better retention of hydrophilic peptides.
Chromatographic profile of peptides obtained on trypsin digestion of adalimumab using HPLC for peptide analysis
Figure 4.Base peak chromatogram of adalimumab tryptic peptides. Heavy chain glycosylated peptide (EEQYN301STYR) is observed at 24.03 min. Using a dual column set up, we observe hydrophilic peptides such as PGK, SCDK, VDK eluting at under 4 min. An example peptide and its oxidized form are NSLYLQM83NSLR and NSLYLQMNSLR eluting at 49.30 and 64.16 min, respectively.
Peptide Mapping Method Conditions
Columns:Two coupled Ascentis® Express Peptide ES-C18 columns 15 cm x 1.0 mm, 2.7 μm (53561-U)
Mobile phase:[A] 0.1% formic acid (FA) in water; [B] 0.1% FA in acetonitrile (v/v)
Gradient:Time (min)A (%)B (%)
 0.00991
 1206535
 121397
 136397
 137991
 162991
Flow:0.08 mL/min
Column Temp.:Ambient
Detection:MS/MS
Injection:10 µL
Table 2.Peptide mapping method conditions
The depth of information provided by peptide mapping has led to the development of multi-attribute methods (MAM) that are used to ensure quality and consistency of protein therapeutics with a single method.13 With MAMs, a list of PQAs (product quality attributes) is generated from an in-depth characterization of the protein-of-interest using high-resolution, high-mass accuracy MS/MS, and peptide mapping experiments. These analyses include characterization of PTMs, particularly glycosylation. Results of the in-depth characterization are used to create a library of features expected from the therapeutic products during their manufacture. A simpler, MS only instrument can then be used to monitor the essential PQAs, or CQAs, during on-going monitoring of drug manufacture. The occurrence of newly identified, missing, or changed peaks during analysis triggers further investigation. The mild, rapid digestion provided by FASP and the low artifact digestion buffer, along with the excellent sequence coverage here, lends itself to MAM quality control of protein therapeutics.
N-Linked Glycan Analysis
A prerequisite for accurate quantification of glycans is their complete release and isolation from the protein. Enzymatic cleavage with PNGase F is remarkably effective for achieving this. PNGase F is characterized by its high activity, broad substrate specificity, and its property of cutting most asparagine linked (N-linked) glycans. The glycan residue remains intact and can be subjected to further analysis. Here, a PNGase Fast Kit was used which, in combination with the enzyme, provided drastically reduced reaction time but with an accuracy equivalent to traditional methods.
Among glycan derivatization reagents, procainamide offers better fluorescence and electrospray ionization, and therefore better sensitivity than the more traditional 2-AB and 2-AA labeling. Here, released N-glycans were analyzed by a UHPLC-FLR-MS method that takes advantage of the quantitation provided by fluorescence detection and mass spectrometric identification. In total, 16 glycan features of adalimumab were observed. Figure 5 illustrates the fluorescence chromatogram of adalimumab glycans. while Table 3 provides the chromatographic conditions. Glycan peak areas were integrated, and the MS spectra were used to confirm glycan identities (Table 4). In total, twelve different glycans were quantified using this approach. The glycan profile, including qualitative and quantitative aspects, is comparable to the results obtained elsewhere.14,15
Fluorescence chromatogram of adalimumab after cleavage using PNGase and derivatization with procainamide for glycan analysis Keywords: Glycan analysis, glycan profiling, glycan profile, monoclonal antibody characterization
Figure 5.Fluorescence chromatogram of adalimumab after PNGase release and derivatization with procainamide. Peak annotations correspond to glycans shown in Table 4.
Glycan Analysis Method Conditions
Column:BIOshell™ Glycan 15 cm x 2.1 mm, 2.7 μm (50994-U)
Mobile phase:[A] 75 mM ammonium formate, pH 4.4 in water; [B] acetonitrile
Gradient:Time (min)A (%)B (%)
 0.007525
 755941
 767525
Flow rate:0.3 mL/Min
Column Temp.:58° C
Detection:1:1 split to both fluorescence detector, Ex: 308 nm, Em: 359 nm, and MS
Injection:10 µL
Table 3.Glycan analysis method conditions
Peak #GlycanMonosaccaride CompositionContent (%)
1G0-NMan3 GlcNAc30.6
2G0F-NFuc1 Man3 GlcNAc34.1
3G0Man3 GlcNAc40.9
4Man5Man5 GlcNAc25.3
5G0FFuc1 Man3 GlcNAc465.0
6G1F-NFuc1 Man3 Gal1 GlcNAc31.7
7aG1(1,6)Man3 Gal1 GlcNAc40.4
7bG1(1,3)Man3 Gal1 GlcNAc40.4
8Man6Man6 GlcNAc21.8
9aG1F(1,6)Fuc1 Man3 Gal1 GlcNAc412.1
9bG1F(1,3)Fuc1 Man3 Gal1 GlcNAc45.1
10G2FFuc1 Man3 Gal2 GlcNAc42.7
Table 4.N-Glycans, monosaccharide composition, and content in adalimumab
Conclusion
Characterization and monitoring of therapeutic mAbs is required by regulatory authorities to ensure efficacy and safety of these drugs. Workflows for three important approaches to mAb analysis were developed and presented elsewhere as detailed step-by-step procedures.(7-9)
See our application collection for Biopharmaceutical Characterization at our Pharma QC page SigmaAldrich.com/PharmaQC
Columns
Sorry, an unexpected error has occurred
Network error: Failed to fetch
System Suitability Standards and Samples
Sorry, an unexpected error has occurred
Network error: Failed to fetch
Sample Preparation
Sorry, an unexpected error has occurred
Network error: Failed to fetch
Other Solvents and Reagents
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
Berkowitz SA, Engen JR, Mazzeo JR, Jones GB. 2012. Analytical tools for characterizing biopharmaceuticals and the implications for biosimilars. Nat Rev Drug Discov. 11(7):527-540. https://doi.org/10.1038/nrd3746
2.
Song YE, Dubois H, Hoffmann M, D?Eri S, Fromentin Y, Wiesner J, Pfenninger A, Clavier S, Pieper A, Duhau L, et al. 2021. Automated mass spectrometry multi-attribute method analyses for process development and characterization of mAbs. Journal of Chromatography B. 1166122540. https://doi.org/10.1016/j.jchromb.2021.122540
3.
Knox JH. 1978. High Performance Liquid Chromatography. Edinburgh University Press..
4.
D'Atri V, Fekete S, Beck A, Lauber M, Guillarme D. 2017. Hydrophilic Interaction Chromatography Hyphenated with Mass Spectrometry: A Powerful Analytical Tool for the Comparison of Originator and Biosimilar Therapeutic Monoclonal Antibodies at the Middle-up Level of Analysis. Anal. Chem.. 89(3):2086-2092. https://doi.org/10.1021/acs.analchem.6b04726
5.
Sjögren J, Olsson F, Beck A. 2016. Rapid and improved characterization of therapeutic antibodies and antibody related products using IdeS digestion and subunit analysis. Analyst. 141(11):3114-3125. https://doi.org/10.1039/c6an00071a
6.
Beck A, Wagner-Rousset E, Ayoub D, Van Dorsselaer A, Sanglier-Cianférani S. 2013. Characterization of Therapeutic Antibodies and Related Products. Anal. Chem.. 85(2):715-736. https://doi.org/10.1021/ac3032355
7.
Altmaier S, Jalili P, Sreenivasan U, Ray K. Protocol for Purification, Optional Reduction, and SEC-MS Analysis of a Monoclonal Antibody. [Internet].[cited 21 Mar 2022]. Available from: /AT/en/technical-documents/protocol/pharmaceutical-and-biopharmaceutical-manufacturing/biologics-biosimilars-characterization/protocol-for-analysis-of-a-monoclonal-antibody
8.
Rule G, Jalili P, Sreenivasan U, Ray K. Peptide Mapping LC- MS/MS Analysis Workflow for Adalimumab. [Internet].[cited 21 Mar 2022]. Available from: /AT/en/technical-documents/protocol/pharmaceutical-and-biopharmaceutical-manufacturing/biologics-biosimilars-characterization/peptide-mapping-lc-ms-ms-analysis-workflow-for-adalimumab
9.
Rigger R, Jalili P, Sreenivasan U, Ray K. Step-by-Step Protocol for Procainamide Labelled Glycan Profiling of a Monoclonal Antibody. [Internet].[cited 21 Mar 2022]. Available from: /AT/en/technical-documents/protocol/pharmaceutical-and-biopharmaceutical-manufacturing/biologics-biosimilars-characterization/released-n-linked-glycan-analysis-workflow
10.
Lermyte F, Tsybin YO, O'Connor PB, Loo JA. 2019. Top or Middle? Up or Down? Toward a Standard Lexicon for Protein Top-Down and Allied Mass Spectrometry Approaches. J. Am. Soc. Mass Spectrom.. 30(7):1149-1157. https://doi.org/10.1007/s13361-019-02201-x
11.
Jalili P, Ray K, Turner J, Dube M. 2021. LCGC Europe ebook,. 3-12.
12.
Wiśniewski JR, Zougman A, Nagaraj N, Mann M. 2009. Universal sample preparation method for proteome analysis. Nat Methods. 6(5):359-362. https://doi.org/10.1038/nmeth.1322
13.
Rogers RS, Nightlinger NS, Livingston B, Campbell P, Bailey R, Balland A. 2015. Development of a quantitative mass spectrometry multi-attribute method for characterization, quality control testing and disposition of biologics. mAbs. 7(5):881-890. https://doi.org/10.1080/19420862.2015.1069454
14.
Lee N, Lee JJ, Yang H, Baek S, Kim S, Kim S, Lee T, Song D, Park G. 2019. Evaluation of similar quality attribute characteristics in SB5 and reference product of adalimumab. mAbs. 11(1):129-144. https://doi.org/10.1080/19420862.2018.1530920
15.
Tebbey PW, Varga A, Naill M, Clewell J, Venema J. 2015. Consistency of quality attributes for the glycosylated monoclonal antibody Humira® (adalimumab). mAbs. 7(5):805-811. https://doi.org/10.1080/19420862.2015.1073429