Merck
HomeDNA & RNA PurificationMammalian Genomic DNA Miniprep Kit
Mammalian Genomic DNA Miniprep Kit
For purification of genomic DNA from a variety of mammalian sources
Description: The GenElute Mammalian Genomic DNA Purification Kit provides a simple and convenient way to isolate pure, high molecular weight DNA from a variety of mammalian sources (Figure 1 and Figure 2). These kits use a silica-based membrane, specially selected for genomic DNA purification, in a convenient spin column format (G1N10G1N70G1N350).
Procedure
Mammalian cells and tissues are lysed with a chaotropic salt-containing buffer to ensure denaturation of macromolecules. DNA is bound to the spin column membrane and the remaining lysate is removed by centrifugation. A filtration column is used to remove cell debris. After washing to remove contaminants, the DNA is eluted with buffer into a collection tube. The purified DNA may be used in many applications such as sequencing, cloning, blotting, restriction digestion, ligation, and PCR.
Features and Benefits
  • Typical DNA yields of 25 µg from 2 x 106 cultured cells or 30 µg from 25 mg of tissue (Figure 4)
  • Preparation time is only 20 minutes after lysis
  • Purified genomic DNA has A260/A280 ratios between 1.6 and 1.9
  • No need for mechanical homogenization
  • 40% more purification preps offered than other leading supplier
Storage
Room Temperature
R: 20/21/22 S: 26-36
Application
Purified DNA may be used in many applications such as sequencing, cloning, blotting, restriction digestion (Figure 3), ligation, and PCR (Figure 4).
GenElute Mammalian Genomic Flow Chart
MaterialAmountTypical Yield
Jurkat Cells (human)2 x 106 Cells5 - 10 µg
HEK293 Cells (human)2 x 106 Cells10 - 20 µg
HeLa Cells (human)2 x 106 Cells15 - 25 µg
Mouse Pancreas Tissue
20 mg10 - 25 µg
Mouse Spleen Tissue10 mg10 - 25 µg
Mouse Thymus Tissue16 mg10 - 25 µg
Mouse Lung Tissue20 mg5 - 15 µg
Mouse Brain Tissue16 mg5 - 15 µg
Mouse Kidney Tissue20 mg10 - 25 µg
Mouse Liver Tissue25 mg10 - 30 µg
Table 1. Yields produced using GenElute Mammalian Genomic DNA Purification Kits (with RNase and Proteinase K treatment).
Genomic DNA Purified from
Mammalian Tissues
Genomic DNA purified from mammalian tissues
Figure 1. Genomic DNA purified from mammalian tissues.Purified genomic DNA, from the indicated mouse tissues, was isolated with the GenElute™ Mammalian Genomic DNA Miniprep Kit. Genomic DNA (200 ng/lane) was analyzed on a 0.8% agarose gel to illustrate yield and integrity. Markers are lambda DNA digested with Hind II. (Product No. D9780)
Genomic DNA Purified from Cells using GenElute Mammalian Genomic DNA Purification Kit
Genomic DNA purified from mammalian cells
Figure 2. Genomic DNA purified from mammalian cells.Purified genomic DNA was isolated with the GenElute Mammalian Genomic DNA Purification Kit from 2 x 106 cells of the sources indicated. The genomic DNA (200 ng/lane) was analyzed on a 0.8% agarose gel. Marker is a Lambda Hind III digest (0.5-23 kb).
Genomic DNA Purified with GenElute Mammalian Genomic DNA Purification Kit is Suitable for Restriction Enzyme Digestions.
 Mammalian Restriction Enzyme
Figure 3.Genomic DNA (200 ng) was digested with EcoR I (10 units in 20 µL at 37°C for 4 hours) followed by electrophoresis (150 ng/lane) on a 0.8% agarose gel. The undigested DNA was incubated under the same conditions, but without EcoR I. Marker is a Lambda Hind III digest (0.5-23 kb).
Yield Comparison
GenElute Mammalian Genomic Yield
Figure 4.Yields of genomic DNA compared with leading Supplier Q using 2 x 200 µL elutions.
Order Information
Sorry, an unexpected error has occurred
Network error: Failed to fetch