Merck
HomeMammalian Cell CultureCoating Tissue Culture Plates with Vitronectin
Coating Tissue Culture Plates with Vitronectin
Vitronectin (Product No. SRP3186) is used to coat culture vessels for cells attachment. Below is a suggested coating protocol for a 96-well plate at a coating concentration of 0.5 µg/cm2. Note that the optimal concentration will vary with each cell line. For culture vessels other than a 96-well plate, scale this protocol accordingly. To calculate the working concentration of vitronectin, use Table 1, or the following equation:
Working concentration calculation
Working concentration calculation Example:
To coat a 96-well plate (surface area is 0.3 cm2/well) at a coating concentration of 0.5 µg/cm2, prepare 9.6-10 mL of diluted vitronectin solution (0.1 mL/well × 96 wells, Table 1) at the following concentration:
Working concentration calculation2
The vitronectin working concentration in this example is 1.5 µg/mL. Adding 0.1 mL per well of this working solution will yield a coating concentration of 0.5 µg/cm2.
Bioassay
Purity
Bioassay
The biological activity of human, recombinant Vitronectin (VTN) expressed in HEK 293 cells (SRP3186) was measured in a cell attachment assay using CHO cells. The EC50 is defined as the effective concentration that elicits a 50% increase in cell attachment in a cell-based bioassay.
Purity
The purity of human, recombinant vitronectin expressed in HEK 293 cells (SRP3186) was assessed on SDS-PAGE (1.25 μg/lane.) followed by InstantBlue™ stain (ISB1L). (Representative result)
Materials
  • 1X PBS
  • Pure water (e.g.,Product No. W3500)
  • Sterile tissue culture 96-well plate with a lid (Product No. CLS3300)
  • Recombinant human vitronectin (Product No. SRP3186)
Protocol
A. Preparation of vitronectin
  1. Reconstitute the lyophilized vitronectin with sterile water, to a final concentration of 100-300 µg/mL. This stock solution can be stored at 2-8 oC for up to one week. For extended storage, it is best to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20 to -80 oC.
  2. Further dilute the reconstituted vitronectin stock solution in 1X PBS to a final concentration of 1.5 µg/mL. For example, for a 10 mL solution (sufficient to coat one 96-well plate), dilute 75 µL of 200 µg/mL stock solution in 10 mL of 1X PBS.
B. Coating the plate
  1. To each well, add 100 µL of the 1.5 µg/mL vitronectin solution, for a final coating concentration of 0.5 µg/cm2.
  2. Cover the plate with a lid. For best results it is best to incubate the plate for 2 hours at 37 oC in 5% CO2 and 95% air, and then over-night at 2-8 oC.
  3. Aspirate the vitronectin solution, being careful not to scratch the coated vitronectin. The plate is now ready to use.
  4. Wash each well 3 times with 1XPBS.
  5. For immediate use, aspirate the 1XPBS. Alternatively, the coated plate can be stored for up to one week at 2-8 oC with 1XPBS, covered and wrapped in laboratory film.
  6. Before use, pre-warm the coated plate.
Culture DishApprox. surface areaDiluted vitronectin volumeVitronectin working concentration
for 0.1 µg/cm2for 0.5 µg/cm2for 1 µg/cm2
96-well plate0.3 cm20.1 mL/well0.3 µg/mL1.5 µg/mL3 µg/mL
24-well plate2 cm20.2 mL/well1 µg/mL5 µg/mL10 µg/mL
12-well plate4 cm20.4 mL/well1 µg/mL5 µg/mL10 µg/mL
6-well plate9 cm21 mL/well0.9 µg/mL4.5 µg/mL9 µg/mL
35 mm plate9 cm21 mL0.9 µg/mL4.5 µg/mL9 µg/mL
60 mm plate21 cm21 mL2.1 µg/mL10.5 µg/mL21 µg/mL
100 mm plate55 cm22 mL2.75 µg/mL13.75 µg/mL27.5 µg/mL
150 mm plate152 cm26 mL2.53 µg/mL12.67 µg/mL25.33 µg/mL
T-25 flask25 cm22.5 mL1 µg/mL5 µg/mL10 µg/mL
T-75 flask75 cm27.5 mL1 µg/mL5 µg/mL10 µg/mL
Table 1Volumes and working concentrations of vitronectin for various tissue culture vessels
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch