Merck
USP Analysis of Actos (Pioglitazone) using Ascentis® Express C18
USP Analysis of Actos (Pioglitazone) using Ascentis® Express C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 5 μm HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
mobile phase component
Product No.
Description
Pricing
Ammonium acetate
LiChropur, eluent additive for LC-MS
standard
Product No.
Description
Pricing
Benzophenone
ReagentPlus®, 99%
Pioglitazone hydrochloride
United States Pharmacopeia (USP) Reference Standard
CONDITIONS
column
Ascentis® Express C18, 15 cm x 4.6 mm, 5 μm particles (50537-U)
mobile phase
[A] 100 mM ammonium acetate; [B] acetonitrile; [C] acetic acid; (55:45:2, A:B:C)
flow rate
0.7 mL/min
pressure
1160 psi (80 bar)
column temp.
25 °C
detector
UV, 269 nm
injection
20 μL
sample
pioglitazone 50 μg/mL, benzophenone 13 μg/mL in mobile phase
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany