Merck
LC/MS Analysis of 2C Amines Drugs of Abuse on Ascentis® Express HILIC
LC/MS Analysis of 2C Amines Drugs of Abuse on Ascentis® Express HILIC application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express HILIC, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
mobile phase component
Product No.
Description
Pricing
Ammonium formate
eluent additive for LC-MS, LiChropur, ≥99.0%
standard
Product No.
Description
Pricing
4-Bromo-2,5-dimethoxyphenethylamine hydrochloride solution
1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
2C-B-FLY hydrochloride solution
1 mg/mL in acetonitrile: water (9:1), ampule of 1 mL, certified reference material, Cerilliant®
CONDITIONS
column
Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 μm particles (53939-U)
mobile phase
5 mM ammonium formate in acetonitrile:water (90:10, v/v)
flow rate
0.4 mL/min
column temp.
35 °C
detector
TOF/MS
detector
2.0 μL
sample
300 ng/mL in acetonitrile
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany