Merck
LC/MS Analysis of 25-Hydroxyvitam D3 and 25-Hydroxyvitamin D2 on Ascentis® Express C18
LC/MS Analysis of 25-Hydroxyvitam D3 and 25-Hydroxyvitamin D2 on Ascentis® Express C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 5 cm × 2.1 mm
standard
Product No.
Description
Pricing
25-Hydroxycholecalciferol
≥98% (HPLC)
25-Hydroxyvitamin D2
≥95.0% (HPLC)
CONDITIONS
column
Ascentis Express C18, 5 cm x 2.1 mm I.D., 2.7 μm particles (53822-U)
mobile phase
[A] 10mM ammonium formate in water; [B] 10mM ammonium formate in acetonitrile; (32:68, A:B)
flow rate
0.4 mL/min
pressure
1711 psi (118 bar)
column temp.
35 °C
detector
ESI(+)
injection
2 μL
sample
10 μg/mL in 25:75, water:methanol
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany