Merck
HPLC Analysis of Proteins on BIOshell A400 Protein C18, Hydrophobicity Comparision
HPLC Analysis of Proteins on BIOshell A400 Protein C18, Hydrophobicity Comparision application for HPLC
Materials
analytical column
Product No.
Description
Pricing
BIOshell A400 Protein C18 Column
L × I.D. 10 cm × 2.1 mm, 3.4 μm particle size
standard
Product No.
Description
Pricing
Superoxide Dismutase from bovine erythrocytes
BioReagent, ≥3,000 units/mg protein, lyophilized powder, suitable for cell culture
Carbonic Anhydrase from bovine erythrocytes
lyophilized powder, ≥2,000 W-A units/mg protein
Catalase from bovine liver
lyophilized powder, ≥10,000 units/mg protein
β-Amylase from sweet potato
β-Lactoglobulin A from bovine milk
≥90% (PAGE)
Bovine Serum Albumin
heat shock fraction, pH 7, ≥98%
α-Lactalbumin from bovine milk
Type III, calcium depleted, ≥85% (PAGE), lyophilized powder
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides
BioUltra, recombinant, expressed in E. coli, ammonium sulfate suspension, ≥95% (SDS-PAGE), ≥550 units/mg protein (biuret)
CONDITIONS
column
BIOShell A400 C18, 10 cm x 2.1 mm I.D., 3.4 μm particles (67463-U)
mobile phase
[A] (water, 0.1% TFA); (acetonitrile, 0.08% TFA), 75:25; [B] (water, 0.1% TFA); (acetonitrile, 0.08% TFA), 25:75
gradient
10 to 100% B in 18 min
flow rate
0.3 mL/min
pressure
2800 psi total pressure (193 bar)
column temp.
30 °C
detector
UV, 215 nm
injection
5 μL
sample
superoxide dismutase, 0.13; a-lactalbumin, 0.13; serum albumin bovine, 0.13; ß-lactoglobulin A, 0.06; catalase, 0.13; (carbonic anhydrase, 0.13; ß-amylase, 0.13; glucose-6 phosphate dehydrogenase, 0.13 g/L in mobile phase A)
Description
Analysis Note
Analysis of proteins of various hydrophobicity on BIOshell A400 Protein C18 to compare the selectivity and retention times.
Legal Information
null