HPLC Analysis of Mianserin Enantiomers on Astec® Cellulose DMP, LC/MS Conditions
![HPLC Analysis of Mianserin Enantiomers on Astec® Cellulose DMP, LC/MS Conditions application for HPLC](/deepweb/assets/sigmaaldrich/product/images/649/275/e2b0a664-c5f4-4ec7-bf72-3fb52dd87af7/800/e2b0a664-c5f4-4ec7-bf72-3fb52dd87af7.jpg)
CONDITIONS
column
Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 μm particles (51098AST)
mobile phase
[A] methanol; [B] ammonium formate; (100:0.1, A:B)
flow rate
0.5 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
2 μL
sample
2 mg/mL in mobile phase