HPLC Analysis of Flavanone Enantiomers on Astec® CYCLOBOND I 2000 DMP, IPA:Heptane Mobile Phase
![HPLC Analysis of Flavanone Enantiomers on Astec® CYCLOBOND I 2000 DMP, IPA:Heptane Mobile Phase application for HPLC](/deepweb/assets/sigmaaldrich/product/images/199/637/69bd512a-db53-4451-88a3-4fa529e5066e/800/69bd512a-db53-4451-88a3-4fa529e5066e.jpg)
CONDITIONS
column
CYCLOBOND I 2000 DMP, 25 cm x 4.6 mm I.D., 5 μm particles (20724AST)
mobile phase
[A] isopropanol; [B] heptane; (30:70, A:B)
flow rate
0.6 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
5 μL
sample
flavanone, 1 mg/mL in heptane
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null