Merck
HPLC Analysis of Erythropoietin on SRT® SEC-300 Gel Filtration Column
HPLC Analysis of Erythropoietin on SRT® SEC-300 Gel Filtration Column application for HPLC
Materials
analytical column
Product No.
Description
Pricing
SRT® SEC-300 Gel Filtration Column, 5 μm
L × I.D. 30 cm × 7.8 mm, 5 μm particle size, stainless steel
mobile phase component
Product No.
Description
Pricing
Sodium phosphate dibasic
99.95% trace metals basis
CONDITIONS
column
SRT SEC-300, 30 cm x 7.8 mm I.D., 5 μm partices, 300 Å, (Z777051)
mobile phase
Na2HPO4-KH2PO4-NaCl
flow rate
0.7 mL/min
column temp.
Ambient
detector
UV, 280 nm
injection
20 μL
sample
EPO (Recombinant Human Erythropoietin)
Description
Analysis Note
Erythropoietin (EPO) is a glycoprotein hormone that controls red blood cell production. EPO was one of the first genetically engineered therapeutic proteins. It is available under such brand names as Epogen and Aranesp for the treatment of anemia. Human EPO has a molecular mass of 34 kDa. The 5 micron SRT SEC-300 column is operated at a flow rate of 0.7 mL/min. Note that after the main peak, an impurity elutes at about 16 minutes.
Legal Information
SRT is a registered trademark of Sepax Technologies