HPLC Analysis of Antihypertensive and Diuretic Drugs on Discovery® RP-Amide C16
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery RP-AmideC16, 15 cm × 4.6 mm I.D., 5 μm particles (505013)
mobile phase
[A] methanol; [B] 25 mM potassium phosphate, dibasic, pH 7.0 (22:78, A:B)
flow rate
1 mL/min
column temp.
30 °C
detector
UV, 254 nm
injection
10 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany