HPLC Analysis of Antihypertensive ACE Inhibitors on Discovery® RP-AmideC16
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery RP-AmideC16, 15 cm × 4.6 mm I.D., 5 μm particles (505013)
mobile phase
[A] acetonitrile; [B] 25 mM potassium phosphate, pH 2.3 (33:67, A:B)
flow rate
0.6 mL/min
column temp.
35 °C
detector
UV, 214 nm
injection
3 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany