Merck
HPLC Analysis of an Energy Drink on Ascentis® Express HILIC (ELSD Detection)
HPLC Analysis of an Energy Drink on Ascentis® Express HILIC (ELSD Detection) application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express HILIC, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 5 cm × 3 mm
CONDITIONS
column
Ascentis Express HILIC, 5 cm x 3 mm I.D., 2.7 μm particles (53967-U)
mobile phase
[A] 100 mM ammonium acetate, pH 5 (titrated with acetic acid) [B] water [C] acetonitrile; (9:1:90, A:B:C:)
flow rate
0.6 mL/min
column temp.
35 °C
detector
ELSD, 55° C, 3.5 bar nitrogen
injection
2 μL
sample
Energy Drink Diluted 1:9 in acetonitrile
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany