Merck
GC Analysis of Residual Solvents on Equity-5
GC Analysis of Residual Solvents on Equity-5 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Equity-5 Capillary GC Column
L × I.D. 30 m × 0.53 mm, df 5.00 μm
CONDITIONS
column
Equity-5, 30 m × 0.53 mm I.D., 5.0 μm (28279-U)
oven
40 °C (6 min), 2 °C/min to 100 °C
inj. temp.
225 °C
detector
FID, 250 °C
carrier gas
helium, 20 cm/sec @ 40 °C
injection
1 μL, 10:1 split
liner
split, cup design
sample
5 ng on-column of a 24 component solvent standard in DMSO
Description
Legal Information
null