Merck
GC Analysis of Oxygen, Nitrogen, and Carbon Dioxide on Carboxen®-1010 PLOT
GC Analysis of Oxygen, Nitrogen, and Carbon Dioxide on Carboxen®-1010 PLOT suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Carboxen®-1010 PLOT Capillary GC Column
L × I.D. 30 m × 0.53 mm, average thickness 30 μm
CONDITIONS
column
Carboxen-1010 PLOT, 30 m x 0.53 mm I.D. (25467)
oven
35 °C (7.0 min), 24 °C/min to 225 °C
inj. temp.
200 °C
detector
TCD, 230 °C, Methanizer, 375 °C, and FID, 230 °C in series
carrier gas
helium, 3.0 mL/min
injection
10 μL, direct using a sample valve, 150 °C
sample
Oxygen at 20% and carbon dioxide at 1.0% in nitrogen
Description
Legal Information
Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany