Merck
  • Two methods for the separation of monounsaturated octadecenoic acid isomers.
Two methods for the separation of monounsaturated octadecenoic acid isomers.
Journal of chromatography. A (2009-12-22)
C Villegas, Y Zhao, J M Curtis
ABSTRACT
The identification and quantification of complex mixtures of cis and trans octadecenoic (18:1) fatty acid isomers presents a major challenge for conventional one-dimensional GC/FID analysis of their methyl esters. We have compared the use of two methods to achieve optimized separations of positional and geometrical octadecenoic fatty acid isomers-comprehensive two-dimensional gas chromatography (GCxGC), and silver ion high performance liquid chromatography interfaced to atmospheric pressure photoionization (APPI) mass spectrometry. Nine isomers of octadecenoic acid methyl ester were well separated on a single silver ion column with a mobile phase of 0.018% acetonitrile and 0.18% isopropanol in hexane. Reproducible retention times were obtained with relative standard deviations of around 1% over 5 injections. The extra selectivity and reproducibility afforded by APPI-MS, together with the wide separation of cis and trans isomers by silver ion chromatography, resulted in a promising method for measurement of octadecenoic acid FAME. The GCxGC separation was performed using various column combinations, and optimal separation was obtained by coupling an ionic liquid column (Supelco SLB-IL100 [1,9-di(3-vinyl-imidazolium) nonane bis(trifluoromethyl) sulfonyl imidate]) in the first dimension with a SGE BPX50 (50% phenyl polysilphenylene-siloxane) in the second dimension. These methods have been applied to the analysis of octadecenoic acid in milk and beef fat.
Product Description
Supelco
SLB®-1ms Capillary GC Column, L × I.D. 15 m × 0.32 mm, df 0.25 μm
Supelco
SLB®-1ms Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.25 μm
Supelco
SLB®-1ms Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.10 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.10 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 10 m × 0.10 mm, df 0.10 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 15 m × 0.10 mm, df 0.10 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 0.25 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.50 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 15 m × 0.25 mm, df 0.25 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.25 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 20 m × 0.18 mm, df 0.18 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 1.00 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.25 μm