Merck
  • Development of an Automated Column Solid-Phase Extraction Cleanup of QuEChERS Extracts, Using a Zirconia-Based Sorbent, for Pesticide Residue Analyses by LC-MS/MS.
Development of an Automated Column Solid-Phase Extraction Cleanup of QuEChERS Extracts, Using a Zirconia-Based Sorbent, for Pesticide Residue Analyses by LC-MS/MS.
Journal of agricultural and food chemistry (2015-02-24)
Bruce D Morris, Richard B Schriner
ABSTRACT
A new, automated, high-throughput, mini-column solid-phase extraction (c-SPE) cleanup method for QuEChERS extracts was developed, using a robotic X-Y-Z instrument autosampler, for analysis of pesticide residues in fruits and vegetables by LC-MS/MS. Removal of avocado matrix and recoveries of 263 pesticides and metabolites were studied, using various stationary phase mixtures, including zirconia-based sorbents, and elution with acetonitrile. These experiments allowed selection of a sorbent mixture consisting of zirconia, C18, and carbon-coated silica, that effectively retained avocado matrix but also retained 53 pesticides with <70% recoveries. Addition of MeOH to the elution solvent improved pesticide recoveries from zirconia, as did citrate ions in CEN QuEChERS extracts. Finally, formate buffer in acetonitrile/MeOH (1:1) was required to give >70% recoveries of all 263 pesticides. Analysis of avocado extracts by LC-Q-Orbitrap-MS showed that the method developed was removing >90% of di- and triacylglycerols. The method was validated for 269 pesticides (including homologues and metabolites) in avocado and citrus. Spike recoveries were within 70-120% and 20% RSD for 243 of these analytes in avocado and 254 in citrus, when calibrated against solvent-only standards, indicating effective matrix removal and minimal electrospray ionization suppression.
Product Description
Supelco
Supel QuE, Z-Sep+ Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep+ Bulk, pkg of 20 g
Supelco
Supel QuE, Z-Sep Tube, centrifuge tube volume 2 mL, pack of 100 ea
Supelco
Supel QuE, Z-Sep+ Tube, centrifuge tube volume 2 mL, pack of 100 ea
Supelco
Supel QuE, Z-Sep Bulk
Supelco
Supel QuE, Z-Sep/C18 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep/C18 Tube, centrifuge tube volume 2 mL, pack of 100
Supelco
Supel QuE, Z-Sep Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep+ with MgSO4 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible