Merck
  • Saliva as an analytical tool to measure occupational exposure to toluene.
Saliva as an analytical tool to measure occupational exposure to toluene.
International archives of occupational and environmental health (2008-01-04)
M Ferrari, S Negri, P Zadra, S Ghittori, M Imbriani
ABSTRACT
To describe a sensitive and rapid method for the determination of toluene in saliva. Biomonitoring of toluene exposure is commonly performed by determination of urinary hippuric acid, o-cresol or toluene itself. The analysis of blood toluene has been verified as another method for biomonitoring. However, drawing blood is invasive and can often not be performed at the workplace for hygienic reasons. Sampling of saliva may be non-invasive, easy to perform and a viable alternative for biomonitoring in the workplace. We measured the solvent concentration in saliva specimens of 5 healthy volunteers studied in the laboratory and a group of 36 workers exposed to toluene in the synthetic leather industry. Saliva was collected into Salivette (Sarstedt, Germany) devices by sterile cotton rolls placed in the mouth and then squeezed into pre-weighted vials. Environmental toluene was collected for the duration of a work-shift by Radiello (FSM, Italy) passive samplers. Toluene in urine and saliva (head space analysis) and in environmental samples was measured by GC-MS. Environmental toluene levels ranged from 0.22 to 57.20 mg/m(3), while the concentrations of the solvent in saliva and urine ranged from 0.12 to 18.30 microg/L, and from 0.47 to 26.64 microg/L, respectively. The correlation coefficients (r) between biological and environmental levels of toluene were 0.77 and 0.93, respectively, for saliva and urine samples. This preliminary study suggests that saliva may offer many advantages over 'classical' biological fluids such as blood as it is readily accessible and collectible: therefore saliva toluene may be considered as a possible biomarker of exposure to toluene.
Product Description
radiello Diffusive Bodies, white, configured for general use, pk of 20
Supelco
radiello Aldehyde Calibration Standard, 50 μg/mL each component in acetonitrile, pkg of 10 mL
Supelco
radiello BTEX Calibration Kit (CS2 Desorption)
radiello On-Field Thermometer, Reader, pk of 1
radiello Diffusive Bodies, blue, configured for sampling light sensitive compounds, pk of 20
Supelco
radiello Methylene Blue Calibration Standard for H2S (Hydrogen Sulfide), pkg of 100 mL
radiello Empty Storage Tubes, polypropylene (with stopper), volume 12 mL, pk of 100
radiello Diffusive Bodies, permeative, configured for anaesthetic gases and vapors, pk of 20
radiello BTEX/VOC Starter Kit, CS2 Desorption
radiello BTEX/VOC Starter Kit, Thermal Desorption
radiello Clips, suspending Radiello triangular support plate, pk of 20
radiello On-Field Thermometer, Includes one standard Radiello vertical adapter (RAD122) for each thermometer, configured for standard use, pk of 3
radiello Empty Storage Tubes, glass (with stopper), volume 2.8 mL, pk of 100
radiello Diffusive Bodies, yellow, configured for reduced sampling rates, pk of 20
radiello Filtration Kit, pk of 20
Supelco
radiello BTEX Calibration Kit (Thermal Desorption)
radiello 1,3-Butadiene Starter Kit, Thermal Desorption, pkg of 1 kit (2 RAD141 adsorbing cartridges and barcode labels, 1 RAD1202 yellow diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet)
radiello Hydrogen Sulfide Starter Kit
radiello Replacement Mounting Strips, for use with Radiello Outdoor Shelter, pk of 100
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (CS2 Desorption), matrix SS net (100 mesh, 5.8 mm diam.), activated with activated charcoal (30-50 mesh), pk of 20
radiello Barcode Labels, pk of 198
radiello Cartridge Adsorbents, for sampling Anaesthetic Gases/Vapors, matrix SS net with mix of mol sieve and activated charcoal (30-50 mesh), pk of 20
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (thermal desorption), matrix SS net (3 x 8 μm, 4.8 mm diam.), Carbograph, pk of 20
radiello Triangular Support Plate, pk of 20
radiello Vertical Adapter, threaded for standard use, pk of 20
radiello Anesthetic Gases and Vapor Sampler
radiello Cartridge Adsorbents, for sampling Aldehydes, matrix SS net with 2,4-DNPH coated FLORISIL®, pk of 20
radiello Cartridge Adsorbents, for sampling Ammonia (NH3), matrix microporous PE impregnated with phosphoric acid, pk of 20
radiello Outdoor Shelter, pk of 10