Merck
  • Lethal Neonatal LTBL Associated with Biallelic EARS2 Variants: Case Report and Review of the Reported Neuroradiological Features.
Lethal Neonatal LTBL Associated with Biallelic EARS2 Variants: Case Report and Review of the Reported Neuroradiological Features.
JIMD reports (2016-08-31)
Renata Oliveira, Ewen W Sommerville, Kyle Thompson, Joana Nunes, Angela Pyle, Manuela Grazina, Patrick F Chinnery, Luísa Diogo, Paula Garcia, Robert W Taylor
ABSTRACT
Mitochondrial translation defects are important causes of early onset mitochondrial disease. Although the biochemical (combined respiratory chain deficiency) signature and neuroimaging are usually distinctive, they are not diagnostic as the genetic origin of mitochondrial translation defects is heterogeneous. We report a female child, born at term to non-consanguineous parents, who exhibited global hypotonia, failure to thrive, persistent and progressive hyperlactacidaemia with lactic acidosis, liver dysfunction and encephalopathy and died at the age of 5 months. Brain MRI revealed hypogenesis of the corpus callosum, T2 signal abnormalities in the medulla oblongata, pons, midbrain, thalami, cerebellar white matter, and a lactate peak on MRS. Muscle histochemistry showed cytochrome c oxidase (COX)-deficient and ragged-red fibres, while muscle biochemical studies showed decreased activities of mitochondrial respiratory chain complexes I and IV. Whole exome sequencing (WES) identified biallelic EARS2 (NM_001083614) variants, a previously reported start-loss (c.1>G, p.Met1?) variant and a novel missense (c.184A>T, p.Ile62Phe) variant. Patient fibroblasts and muscle homogenate displayed markedly decreased EARS2 protein levels, although decreased steady-state levels of complex I (NDUFB8) and complex IV (MT-CO1 and MT-CO2) subunits were only observed in muscle. Pathogenic variants in EARS2, encoding mitochondrial glutamyl-tRNA synthetase (mtGluR), are associated with Leukoencephalopathy involving the Thalamus and Brainstem with high Lactate (LTBL), a mitochondrial disorder characterised by a distinctive brain MRI pattern and a biphasic clinical course. We further outline the unique phenotypic spectrum of LTBL and review the neuroradiological features reported in all patients documented in the literature.
Sigma-Aldrich
Anti-EARS2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid