Merck
  • Chemoselective and regiospecific Suzuki coupling on a multisubstituted sp(3)-carbon in 1,1-diborylalkanes at room temperature.
Chemoselective and regiospecific Suzuki coupling on a multisubstituted sp(3)-carbon in 1,1-diborylalkanes at room temperature.
Journal of the American Chemical Society (2010-08-12)
Kohei Endo, Takahiro Ohkubo, Munenao Hirokami, Takanori Shibata
ABSTRACT
The palladium-catalyzed Suzuki-Miyaura cross-coupling on a multisubstituted sp(3)-carbon in 1,1-diborylalkanes was achieved at room temperature. The generation of a monoborate intermediate by virtue of the adjacent B atom could result in the chemoselective coupling reaction under ambient conditions.
Sigma-Aldrich
Bis(tri-tert-butylphosphine)palladium(0)