Merck
  • A key role for Rac and Pak signaling in neutrophil extracellular traps (NETs) formation defines a new potential therapeutic target.
A key role for Rac and Pak signaling in neutrophil extracellular traps (NETs) formation defines a new potential therapeutic target.
American journal of hematology (2017-11-11)
Mathilde Gavillet, Kimberly Martinod, Raffaele Renella, Denisa D Wagner, David A Williams
ABSTRACT
NET formation in mice (NETosis) is supported by reactive oxygen species (ROS) production by NADPH oxidase and histone hypercitrullination by peptidylarginine deiminase 4 (PAD4). Rac1 and Rac2, expressed in polymorphonuclear neutrophils (PMNs), regulate the cytoskeleton, cell shape, adhesion, and migration and are also essential components of the NADPH oxidase complex. We aimed to explore the role of the Rac signaling pathway including the upstream guanosine exchange factor (GEF) activator, Vav, and a downstream effector, the p21-activated kinase, Pak, on NETosis in PMNs using a previously described flow-cytometry-based assay. Rac2
Sigma-Aldrich
IPA-3, ≥95% (HPLC), powder