- WEE1 epigenetically modulates 5-hmC levels by pY37-H2B dependent regulation of
WEE1 epigenetically modulates 5-hmC levels by pY37-H2B dependent regulation of
Oncotarget (2018-01-02)
Nupam P Mahajan, Pavani Malla, Shambhavi Bhagwat, Vasundhara Sharma, Amod Sarnaik, Jongphil Kim, Shari Pilon-Thomas, Jeffery Weber, Kiran Mahajan
PMID29290954
ABSTRACT
Epigenetic signaling networks dynamically regulate gene expression to maintain cellular homeostasis. Previously, we uncovered that WEE1 phosphorylates histone H2B at tyrosine 37 (pY37-H2B) to negatively regulate global histone transcriptional output. Although pY37-H2B is readily detected in cancer cells, its functional role in pathogenesis is not known. Herein, we show that WEE1 deposits the pY37-H2B marks within the tumor suppressor gene, isocitrate dehydrogenase 2 (