Merck
  • Molecular basis of differential 3' splice site sensitivity to anti-tumor drugs targeting U2 snRNP.
Molecular basis of differential 3' splice site sensitivity to anti-tumor drugs targeting U2 snRNP.
Nature communications (2017-12-14)
Luisa Vigevani, André Gohr, Thomas Webb, Manuel Irimia, Juan Valcárcel
ABSTRACT
Several splicing-modulating compounds, including Sudemycins and Spliceostatin A, display anti-tumor properties. Combining transcriptome, bioinformatic and mutagenesis analyses, we delineate sequence determinants of the differential sensitivity of 3' splice sites to these drugs. Sequences 5' from the branch point (BP) region strongly influence drug sensitivity, with additional functional BPs reducing, and BP-like sequences allowing, drug responses. Drug-induced retained introns are typically shorter, displaying higher GC content and weaker polypyrimidine-tracts and BPs. Drug-induced exon skipping preferentially affects shorter alternatively spliced regions with weaker BPs. Remarkably, structurally similar drugs display both common and differential effects on splicing regulation, SSA generally displaying stronger effects on intron retention, and Sudemycins more acute effects on exon skipping. Collectively, our results illustrate how splicing modulation is exquisitely sensitive to the sequence context of 3' splice sites and to small structural differences between drugs.
Sigma-Aldrich
5-Bromouridine, 98%
Sigma-Aldrich
Monoclonal Anti-BrdU antibody produced in mouse, clone BU-33, ascites fluid, Immunohistology Grade