MilliporeSigma
  • DBU-promoted carbonylative synthesis of 1,3-oxathiolan-2-ones from propargylic alcohols with TFBen as the CO source.
DBU-promoted carbonylative synthesis of 1,3-oxathiolan-2-ones from propargylic alcohols with TFBen as the CO source.
Organic & biomolecular chemistry (2018-01-25)
Jun Ying, Chao Zhou, Xiao-Feng Wu
ABSTRACT
A DBU-promoted carbonylative cyclization of propargylic alcohols with sulfur was developed. Various 1,3-oxathiolan-2-ones were produced in 61-98% yields under mild conditions in the absence of metal catalysts. TFBen (benzene-1,3,5-triyl triformate) as an efficient and solid CO surrogate and S