Merck
  • A small-molecule inhibitor shows that pirin regulates migration of melanoma cells.
A small-molecule inhibitor shows that pirin regulates migration of melanoma cells.
Nature chemical biology (2010-08-17)
Isao Miyazaki, Siro Simizu, Hideo Okumura, Satoshi Takagi, Hiroyuki Osada
ABSTRACT
The discovery of small molecules that bind to a specific target and disrupt the function of proteins is an important step in chemical biology, especially for poorly characterized proteins. Human pirin is a nuclear protein of unknown function that is widely expressed in punctate subnuclear structures in human tissues. Here, we report the discovery of a small molecule that binds to pirin. We determined how the small molecule bound to pirin by solving the cocrystal structure. Either knockdown of pirin or treatment with the small molecule inhibited melanoma cell migration. Thus, inhibition of pirin by the small molecule has led to a greater understanding of the function of pirin and represents a new method of studying pirin-mediated signaling pathways.