Merck
  • An iterative statistical approach to the identification of protein phosphorylation motifs from large-scale data sets.
An iterative statistical approach to the identification of protein phosphorylation motifs from large-scale data sets.
Nature biotechnology (2005-11-08)
Daniel Schwartz, Steven P Gygi
ABSTRACT
With the recent exponential increase in protein phosphorylation sites identified by mass spectrometry, a unique opportunity has arisen to understand the motifs surrounding such sites. Here we present an algorithm designed to extract motifs from large data sets of naturally occurring phosphorylation sites. The methodology relies on the intrinsic alignment of phospho-residues and the extraction of motifs through iterative comparison to a dynamic statistical background. Results show the identification of dozens of novel and known phosphorylation motifs from recently published serine, threonine and tyrosine phosphorylation studies. When applied to a linguistic data set to test the versatility of the approach, the algorithm successfully extracted hundreds of language motifs. This method, in addition to shedding light on the consensus sequences of identified and as yet unidentified kinases and modular protein domains, may also eventually be used as a tool to determine potential phosphorylation sites in proteins of interest.
Sigma-Aldrich
MS PhosphoMix 3 Heavy, Phosphopeptide Standard for MS
Sigma-Aldrich
MS PhosphoMix 2 Light, Phosphopeptide Standard for MS
Sigma-Aldrich
MS PhosphoMix 2 Heavy, Phosphopeptide Standard for MS
Sigma-Aldrich
MS PhosphoMix 3 Light, Phosphopeptide Standard for MS
Sigma-Aldrich
MS PhosphoMix 1 Heavy, Phosphopeptide Standard for MS
Sigma-Aldrich
MS PhosphoMix 1 Light, Phosphopeptide Standard for MS