Merck
  • Cryo-EM structures of the human endolysosomal TRPML3 channel in three distinct states.
Cryo-EM structures of the human endolysosomal TRPML3 channel in three distinct states.
Nature structural & molecular biology (2017-11-07)
Xiaoyuan Zhou, Minghui Li, Deyuan Su, Qi Jia, Huan Li, Xueming Li, Jian Yang
ABSTRACT
TRPML3 channels are mainly localized to endolysosomes and play a critical role in the endocytic pathway. Their dysfunction causes deafness and pigmentation defects in mice. TRPML3 activity is inhibited by low endolysosomal pH. Here we present cryo-electron microscopy (cryo-EM) structures of human TRPML3 in the closed, agonist-activated, and low-pH-inhibited states, with resolutions of 4.06, 3.62, and 4.65 Å, respectively. The agonist ML-SA1 lodges between S5 and S6 and opens an S6 gate. A polycystin-mucolipin domain (PMD) forms a luminal cap. S1 extends into this cap, forming a 'gating rod' that connects directly to a luminal pore loop, which undergoes dramatic conformational changes in response to low pH. S2 extends intracellularly and interacts with several intracellular regions to form a 'gating knob'. These unique structural features, combined with the results of electrophysiological studies, indicate a new mechanism by which luminal pH and other physiological modulators such as PIP
Sigma-Aldrich
2,2-Dimethyl-1,3-propanediol, 99%
Sigma-Aldrich
Maltose solution, BioReagent, for molecular biology, ~20% in H2O