Merck
  • Structure of human lysosomal acid α-glucosidase-a guide for the treatment of Pompe disease.
Structure of human lysosomal acid α-glucosidase-a guide for the treatment of Pompe disease.
Nature communications (2017-10-25)
Véronique Roig-Zamboni, Beatrice Cobucci-Ponzano, Roberta Iacono, Maria Carmina Ferrara, Stanley Germany, Yves Bourne, Giancarlo Parenti, Marco Moracci, Gerlind Sulzenbacher
ABSTRACT
Pompe disease, a rare lysosomal storage disease caused by deficiency of the lysosomal acid α-glucosidase (GAA), is characterized by glycogen accumulation, triggering severe secondary cellular damage and resulting in progressive motor handicap and premature death. Numerous disease-causing mutations in the gaa gene have been reported, but the structural effects of the pathological variants were unknown. Here we present the high-resolution crystal structures of recombinant human GAA (rhGAA), the standard care of Pompe disease. These structures portray the unbound form of rhGAA and complexes thereof with active site-directed inhibitors, providing insight into substrate recognition and the molecular framework for the rationalization of the deleterious effects of disease-causing mutations. Furthermore, we report the structure of rhGAA in complex with the allosteric pharmacological chaperone N-acetylcysteine, which reveals the stabilizing function of this chaperone at the structural level.
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)