Merck
  • Soluble CD40L is associated with increased oxidative burst and neutrophil extracellular trap release in Behçet's disease.
Soluble CD40L is associated with increased oxidative burst and neutrophil extracellular trap release in Behçet's disease.
Arthritis research & therapy (2017-10-21)
Sandro Félix Perazzio, Paulo Vitor Soeiro-Pereira, Viviane Cardoso Dos Santos, Marlon Vilela de Brito, Bruno Salu, Maria Luiza Vilela Oliva, Anne Margherite Stevens, Alexandre Wagner Silva de Souza, Hans D Ochs, Troy R Torgerson, Antonio Condino-Neto, Luis Eduardo Coelho Andrade
ABSTRACT
Studies have suggested that soluble factors in plasma from patients with active (aBD) and inactive (iBD) Behçet's disease (BD) stimulate neutrophil function. Soluble CD40 ligand (sCD40L) is an important mediator of inflammation in BD. Its expression and effect on neutrophil oxidative burst and neutrophil extracellular trap (NET) release have not been characterized. In this study, we sought to investigate the role of plasma and the CD40L pathway on NET release and the oxidative burst profile in patients with aBD and iBD. Neutrophils and peripheral blood mononuclear cells (PBMCs) were obtained from patients with aBD (n = 30), patients with iBD (n = 31), and healthy control subjects (HCs; n = 30). sCD40L plasma concentration was determined in individual samples. A pool of plasma for each group was created. In some experiments, plasma pools were treated with recombinant CD40 (rhCD40-muIg) for sCD40L blockade. NET release and H sCD40L plasma levels were significantly higher in patients with iBD (median 17,234, range 2346-19,279 pg/ml) and patients with aBD (median 18,289, range 413-19,883 pg/ml) than in HCs (median 47.5, range 33.7-26.7 pg/ml; p < 0.001). NET release was constitutively increased in BD compared with HC. NET release and H Plasma from patients with aBD exerts a stimulus on NET release and oxidative burst, probably induced by sCD40L.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
CD40 human, recombinant, expressed in E. coli, 0.5 mg protein/mL