Merck
  • Role of bone morphogenetic proteins in sprouting angiogenesis: differential BMP receptor-dependent signaling pathways balance stalk
Role of bone morphogenetic proteins in sprouting angiogenesis: differential BMP receptor-dependent signaling pathways balance stalk
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2017-07-25)
Andreas Benn, Christian Hiepen, Marc Osterland, Christof Schütte, An Zwijsen, Petra Knaus
ABSTRACT
Before the onset of sprouting angiogenesis, the endothelium is prepatterned for the positioning of tip and stalk cells. Both cell identities are not static, as endothelial cells (ECs) constantly compete for the tip cell position in a dynamic fashion. Here, we show that both bone morphogenetic protein 2 (BMP2) and BMP6 are proangiogenic
Sigma-Aldrich
2-(1H-Imidazol-2-yl)pyridine, 97%