Merck
  • Cannabinoid Receptor 1 Participates in Liver Inflammation by Promoting M1 Macrophage Polarization
Cannabinoid Receptor 1 Participates in Liver Inflammation by Promoting M1 Macrophage Polarization
Frontiers in immunology (2017-10-17)
Lei Tian, Weiyang Li, Le Yang, Na Chang, Xiaoting Fan, Xiaofang Ji, Jieshi Xie, Lin Yang, Liying Li
ABSTRACT
Macrophage M1/M2 polarization mediates tissue damage and inflammatory responses. Cannabinoid receptor (CB) 1 participated in liver fibrogenesis by affecting bone marrow (BM)-derived monocytes/macrophages (BMMs) activation. However, the knowledge of whether CB1 is involved in the polarization of BMMs remains limited. Here, we found M1 gene signatures (including CD86, MIP-1β, tumor necrosis factor, IL-6, and inducible nitric oxide synthase) and the amount of M1 macrophages (CD86 CB1 plays a crucial role in regulating M1 polarization of BMMs in liver injury, depending on two independent signaling pathways: G(α)
Sigma-Aldrich
MISSION® esiRNA, targeting human CNR2
Sigma-Aldrich
Arachidonyl-2′-chloroethylamide hydrate, ≥97% (HPLC), oil
Sigma-Aldrich
AM281, ≥98% (HPLC)
Sigma-Aldrich
AM630, ≥90% (HPLC)