Merck
  • Efficient coproduction of gluconic acid and xylonic acid from lignocellulosic hydrolysate by Zn(II)-selective inhibition on whole-cell catalysis by Gluconobacter oxydans.
Efficient coproduction of gluconic acid and xylonic acid from lignocellulosic hydrolysate by Zn(II)-selective inhibition on whole-cell catalysis by Gluconobacter oxydans.
Bioresource technology (2017-07-21)
Xuelian Zhou, Xin Zhou, Lu Huang, Rou Cao, Yong Xu
ABSTRACT
With Zn(II)-selective inhibition on the whole-cell catalysis of Gluconobacter oxydans NL71, gluconic acid and xylonic acid were coproduced efficiently from the hydrolysate of corn stover. Further metabolism of gluconic acid to the by-product 2-ketogluconic acid was prevented by addition of 10g/L ZnCl
Sigma-Aldrich
Cellulase from Trichoderma reesei, aqueous solution, ≥700 units/g