- Synthesis and interconversions of reduced, alkali-metal supported iron-sulfur-carbonyl complexes.
Synthesis and interconversions of reduced, alkali-metal supported iron-sulfur-carbonyl complexes.
Dalton transactions (Cambridge, England : 2003) (2017-07-05)
J Patrick Shupp, Amber R Rose, Michael J Rose
PMID28675227
ABSTRACT
We report the synthesis, interconversions and X-ray structures of a set of [mFe-nS]-type carbonyl clusters (where S = S