Merck
  • Mechanisms of Skin Toxicity Associated with Metabotropic Glutamate Receptor 5 Negative Allosteric Modulators.
Mechanisms of Skin Toxicity Associated with Metabotropic Glutamate Receptor 5 Negative Allosteric Modulators.
Cell chemical biology (2017-07-04)
Falgun Shah, Antonia F Stepan, Alison O'Mahony, Sharlene Velichko, Alexandra E Folias, Christopher Houle, Christopher L Shaffer, John Marcek, Jessica Whritenour, Robert Stanton, Ellen L Berg
ABSTRACT
Cutaneous reactions represent one of the most common adverse drug effects observed in clinical trials leading to substantial compound attrition. Three negative allosteric modulators (NAMs) of metabotropic glutamate receptors (mGluRs), which represent an important target for neurological diseases, developed by Pfizer, were recently failed in preclinical development due to delayed type IV skin hypersensitivity observed in non-human primates (NHPs). Here we employed large-scale phenotypic profiling in standardized panels of human primary cell/co-culture systems to characterize the skin toxicity mechanism(s) of mGluR5 NAMs from two different series. Investigation of a database of chemicals tested in these systems and transcriptional profiling suggested that the mechanism of toxicity may involve modulation of nuclear receptor targets RAR/RXR, and/or VDR with AhR antagonism. The studies reported here demonstrate how phenotypic profiling of preclinical drug candidates using human primary cells can provide insights into the mechanisms of toxicity and inform early drug discovery and development campaigns.
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype enteritidis, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Posaconazole, VETRANAL®, analytical standard
Sigma-Aldrich
Anti-Urokinase Antibody, NT fragment, clone 10D.7.2, clone 10D.7.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-tPA (Tissue Plasminogen Activator) Antibody, clone GMA-043, clone GMA-043, Upstate®, from mouse
Sigma-Aldrich
Tpl2 Kinase Inhibitor, The Tpl2 Kinase Inhibitor, also referenced under CAS 871307-18-5, controls the biological activity of Tpl2 Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Anti-CD54 Antibody, extracellular, clone 84H10, clone 84H10, Chemicon®, from mouse
Sigma-Aldrich
Anti-Tissue Factor Mouse mAb (TF9-10H10), lyophilized, clone TF9-10H10, Calbiochem®
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
2-Chloroethyl ethyl sulfide, 97%
Sigma-Aldrich
Anti-PAI-1 (Ab-1) Mouse mAb (2), lyophilized, clone 2, Calbiochem®
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Pomalidomide, ≥98% (HPLC)