Merck
  • Tongue and lingual salivary glands of the canary: scanning electron microscopy and histochemical study.
Tongue and lingual salivary glands of the canary: scanning electron microscopy and histochemical study.
Folia morphologica (2017-02-16)
F Başak, Ş H Atalgin, E Ü Bozkurt
ABSTRACT
In this study, morphological characteristics of the canary tongue were examined macroscopically and histologically besides using scanning electron microscopy. Furthermore, histochemical features of the lingual salivary glands of the canary were also examined. The results suggest that the tongue of the canary has an equilateral quadrangle shape is sloped towards the apex on its dorsal surface; where its sides are bounded by tall epithelial extensions. Additionally, histological examination showed that salivary glands were only present on the body of the tongue and there were no taste buds. However, the tongue has mechanical sen-sory cell groups in its subepithelial connective tissue. Histochemical examination, demonstrated that the salivary gland epithelial cells contained carbohydrates which were composed of acidic sialo-mucins.
Sigma-Aldrich
Periodic Acid-Schiff (PAS) Staining System
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
Sigma-Aldrich
Diastase from Aspergillus oryzae, powder, white, amylase ≥3500 U/g
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid