Merck
  • Polar Lipids Analysis of Cultured Phytoplankton Reveals Significant Inter-taxa Changes, Low Influence of Growth Stage, and Usefulness in Chemotaxonomy.
Polar Lipids Analysis of Cultured Phytoplankton Reveals Significant Inter-taxa Changes, Low Influence of Growth Stage, and Usefulness in Chemotaxonomy.
Microbial ecology (2016-11-12)
José Pedro Cañavate, Isabel Armada, Ismael Hachero-Cruzado
ABSTRACT
The high lipid diversity of microalgae has been used to taxonomically differentiate phytoplankton taxa at the class level. However, important lipids such as phospholipids (PL) and betaine lipids (BL) with potential chemotaxonomy application in phytoplankton ecology have been scarcely studied. The chemotaxonomy value of PL and BL depends on their intraspecific extent of variation as microalgae respond to external changing factors. To determine such effects, lipid class changes occurring at different growth stages in 15 microalgae from ten different classes were analyzed. BL occurred in 14 species and were the less affected lipids by growth stage with diacylglyceryl-hydroxymethyl-N,N,N-trimethyl-b-alanine (DGTA) showing the highest stability. PL were more influenced by growth stage with phosphatidylcholine (PC), phosphatidylglycerol (PG), and phosphatidyletanolamine (PE) declining towards older culture stages in some species. Glycolipids were the more common lipids, and no evident age-related variability pattern could be associated to taxonomic diversity. Selecting BL and PL as descriptor variables optimally distinguished microalgae taxonomic variability at all growth stages. Principal coordinate analysis arranged species through a main tendency from diacylglyceryl-hydroxymethyl-N,N,N-trimethyl-b-alanine (DGCC) containing species (mainly dinoflagellates and haptophytes) to DGTA or PC containing species (mainly cryptophytes). Two diatom classes with similar fatty acid profiles could be distinguished from their respective content in DGTA (Bacillariophyceae) or DGCC (Mediophyceae). In green lineage classes (Trebouxiophyceae, Porphyridophyceae, and Chlorodendrophyceae), PC was a better descriptor than BL. BL and PL explained a higher proportion of microalgae taxonomic variation than did fatty acids and played a complementary role as lipid markers.
Sigma-Aldrich
L-α-Phosphatidylinositol ammonium salt solution from bovine liver, 10 mg/mL in chloroform, ≥98% (TLC)
Sigma-Aldrich
L-α-Phosphatidylcholine, egg yolk, Type XVI-E, ≥99% (TLC), lyophilized powder
Sigma-Aldrich
L-α-Phosphatidyl-DL-glycerol sodium salt from egg yolk lecithin, ≥99% (TLC), lyophilized powder
Sigma-Aldrich
L-α-Phosphatidylethanolamine from egg yolk, Type III, 10 mg/mL in chloroform, ≥97%, solution