- Characteristics and properties of nano-LiCoO
Characteristics and properties of nano-LiCoO
Journal of nanobiotechnology (2017-08-24)
Jean-Pierre Brog, Aurélien Crochet, Joël Seydoux, Martin J D Clift, Benoît Baichette, Sivarajakumar Maharajan, Hana Barosova, Pierre Brodard, Mariana Spodaryk, Andreas Züttel, Barbara Rothen-Rutishauser, Nam Hee Kwon, Katharina M Fromm
PMID28830490
ABSTRACT
LiCoO Several new single source precursors containing lithium (Li Our heterobimetallic single source precursors allow to dramatically reduce the production temperature and time for HT-LCO. The obtained nanoparticles of LiCoO
MATERIALS