Merck
  • Application of Partially Substituted 3,5-Dimethylphenylcarbamate-(3-(2-O-β-Cyclodextrin)-2-Hydroxypropoxy)-Propylsilyl-Appended Silica Particles as Chiral Stationary Phase for Multi-mode High-performance Liquid Chromatography.
Application of Partially Substituted 3,5-Dimethylphenylcarbamate-(3-(2-O-β-Cyclodextrin)-2-Hydroxypropoxy)-Propylsilyl-Appended Silica Particles as Chiral Stationary Phase for Multi-mode High-performance Liquid Chromatography.
Journal of chromatographic science (2017-05-16)
Mingxuan Ma, Qunli Wei, Min Meng, Jiale Yin, Lei Du, Xia Zhu, Mengjun Min, Xueyan Zhou, Xiaoxing Yin, Yinhan Gong
ABSTRACT
A new type of partially substituted 3,5-dimethylphenylcarbamate-(3-(2-O-β-cyclodextrin)-2-hydroxypropoxy)-propylsilyl-appended silica particles (MP-CD-HPS) have been prepared by a convenient post-immobilization derivazition procedure. The MP-CD-HPS has been successfully used as chiral stationary phase (CSP) for high-performance liquid chromatography (HPLC) under normal phase, reversed phase and polar organic mobile phase conditions. The chromatographic evaluation results show that the MP-CD-HPS has excellent selectivity for the separation of aromatic positional isomers and enantiomers of some chiral compounds. The multi-mode HPLC separation results also indicate that both the stable ether spacer linking to the wider torus rim of β-cyclodextrin in the MP-CD-HPS phase and the hydroxyl residues in the partially substituted β-cyclodextrin have important contributions to chiral recognitions and chromatographic separations.
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
3,5-Dimethylphenyl isocyanate, 99%