Merck
  • Ferrocyanide-Mediated Photoreduction of Ferricytochrome C Utilized to Selectively Probe Non-native Conformations Induced by Binding to Cardiolipin-Containing Liposomes.
Ferrocyanide-Mediated Photoreduction of Ferricytochrome C Utilized to Selectively Probe Non-native Conformations Induced by Binding to Cardiolipin-Containing Liposomes.
Chemistry (Weinheim an der Bergstrasse, Germany) (2016-11-20)
Dmitry Malyshka, Reinhard Schweitzer-Stenner
ABSTRACT
Ferricytochrome c binding to cardiolipin-containing liposomes produces a heterogeneous distribution of conformations comprising native-like and non-native misfolded proteins. We utilized the photoreduction of native ferricytochrome c in the presence of potassium ferrocyanide and resonance Raman spectroscopy to probe the population of native and misfolded cytochrome c on liposomes with 20 % tetraoleylcardiolipin (TOCL)/80 % dioleylphosphocholine (DOPC) and with 100 % TOCL as a function of TOCL concentration. Our data provided strong support for an earlier model, which predicts that the equilibrium between native and non-native conformations is shifted to the latter with decreasing protein occupation of liposomes.
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% based on Mol. Wt. 12,384 basis