- Interleukin-18 activates Vγ9Vδ2
Interleukin-18 activates Vγ9Vδ2
Immunology (2017-03-28)
Alanna S Murday, Suchita Chaudhry, C David Pauza
PMID28342224
ABSTRACT
The study aimed to identify an immunoregulatory factor that restores the phosphoantigen response of Vγ9Vδ2
Sigma-Aldrich
Isopentenyl pyrophosphate triammonium salt solution, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)
Pricing and availability is not currently available.